Skip to Content
Merck
All Photos(1)

Key Documents

1.93708

Supelco

Calcium nitrate tetrahydrate

EMPLURA®

Synonym(s):

Calcium nitrate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(NO3)2 · 4 H2O
CAS Number:
Molecular Weight:
236.15
MDL number:
UNSPSC Code:
12352302

product line

EMPLURA®

storage temp.

room temp

InChI

1S/Ca.2HNO3.4H2O/c;2*2-1(3)4;;;;/h;2*(H,2,3,4);4*1H2/q+2;;;;;;

InChI key

DKCJOKYHYIAZQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service