Skip to Content
Merck
All Photos(1)

Key Documents

19-144

Sigma-Aldrich

Phorbol 12-Myristate 13-Acetate

≥99% (HPLC), solid, PKC activator, Upstate®

Synonym(s):

12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, PMA, TPA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
eCl@ss:
32160405
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Phorbol 12-Myristate 13-Acetate,

form

solid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

wet ice

General description

Phorbol 12-Myristate 13-Acetate (PMA) is obtained from the unripe fruit of Sapium indicum. It is a pro-inflammatory agent. PMA shows its effects by the release of cytokines, nicotinamide adenine dinucleotide phosphate (NADPH) oxidases, and proteases that result in tissue damage or activates histamine production. This promotes vascular remodeling, heat, and redness.[1] PMA exhibits antimicrobial, proliferative, antiproliferative, anti-neoplastic effects.[2]

Application

Phorbol 12-Myristate 13-Acetate is suitable as an inducer of cell differentiation in monocytic cell lines such as U937[3] and THP-1.[4][5]

Biochem/physiol Actions

Phorbol 12-Myristate 13-Acetate is a tumor-promoting phorbol ester. It regulates various processes such as gene transcription, cellular growth and differentiation, apoptosis, the immune response, and receptor desensitization via activating protein kinase C (PKC) signaling pathways.[6] It promotes differentiation of human monocytic THP-1 cells to functional macrophages.[5]

Physical form

C36H56O8

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Carc. 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnes Schröder et al.
Journal of orofacial orthopedics = Fortschritte der Kieferorthopadie : Organ/official journal Deutsche Gesellschaft fur Kieferorthopadie (2022-04-22)
Orthodontic tooth movement is a complex process involving the remodeling of extracellular matrix and bone as well as inflammatory processes. During orthodontic treatment, sterile inflammation and mechanical loading favor the production of receptor activator of NF-κB ligand (RANKL). Simultaneously, expression of
Takahide Kohro et al.
Journal of atherosclerosis and thrombosis, 11(2), 88-97 (2004-05-22)
In order to characterize the monocytic cell line THP-1 and its mature, macrophage-like form treated with phorbol 12-myristate 13-acetate (PMA), we have conducted an oligonucleotide microarray assay and compared the results with those from an assay of human monocytes and
12-O-Tetradecanoylphorbol-13-acetate (TPA)-induced increase in depressed white blood cell counts in patients treated with cytotoxic cancer chemotherapeutic drugs.
Han, Z T, et al.
Proceedings of the National Academy of Sciences of the USA, 95, 5362-5365 (1998)
Phorbol 12-myristate 13-acetate upregulates cyclooxygenase-2 expression in human pulmonary epithelial cells via Ras, Raf-1, ERK, and NF-kappaB, but not p38 MAPK, pathways.
Chang, et al.
Cellular Signalling, 17, 299-310 (2018)
J F Valdés López et al.
Morphologie : bulletin de l'Association des anatomistes, 102(338), 205-218 (2018-08-05)
Phorbol-12-myristate-13-acetate (PMA) and 1,25-dihydroxyvitamin D3 (VD3) are stimuli commonly used to induce macrophage differentiation in monocytic cell lines, but the extent of differentiation in comparison to primary tissue macrophages is unclear. Here, we examine the morphological/phenotypic markers associated with differentiation

Questions

  1. What is the solution to reconstitute the PMA

    1 answer
    1. This is soluble in the following solvents as follows:
      DMSO: 100 mg/mL (162.11 mM)
      Ethanol: 100 mg/mL (162.11 mM)
      Water: Insoluble

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service