Skip to Content
Merck
All Photos(2)

Key Documents

19-130

Sigma-Aldrich

Okadaic Acid (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
eCl@ss:
32160405
NACRES:
NA.41
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Quality Level

form

solid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

wet ice

Application

Suitable for use as an inhibitor of serine/threonine-specific protein phosphatases.

Quality

Routinely evaluated as an inhibitor in a kinase assay.

Storage and Stability

Lyophilized: 2 years at 4°C; Rehydrated: 6 months at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Okadaic acid. Protein phosphatase inhibition and muscle contractile effects.
Takai, A
Journal of Muscle Research and Cell Motility, 9, 563-565 (1988)
F M Amara et al.
The Journal of biological chemistry, 271(33), 20126-20131 (1996-08-16)
Mammalian ribonucleotide reductase is a highly regulated activity essential for DNA synthesis and repair. The 3'-untranslated region (3'-UTR) of mammalian ribonucleotide reductase R2 mRNA has been implicated in the tumor promoter, 12-O-tetradecanoylphorbol-13-acetate-mediated stabilization of mouse BALB/c 3T3 R2 message. We
The tumour promoter okadaic acid inhibits reticulocyte-lysate protein synthesis by increasing the net phosphorylation of elongation factor 2.
Redpath, N T and Proud, C G
The Biochemical Journal, 262, 69-75 (1989)
E G Mimnaugh et al.
The Journal of biological chemistry, 270(48), 28654-28659 (1995-12-01)
The abundant, cytoplasmic 90-kDa heat-shock protein associates transiently with the Rous sarcoma virus oncogenic protein tyrosine kinase, pp60v-src, directs its cellular trafficking and negatively regulates its kinase activity. Here we report that the serine/threonine phosphatase inhibitor, okadaic acid, destabilized the
T A Haystead et al.
Nature, 337(6202), 78-81 (1989-01-05)
Okadaic acid is a polyether derivative of 38-carbon fatty acid, and is implicated as the causative agent of diarrhetic shellfish poisoning. It is a potent tumour promoter that is not an activator of protein kinase C, but is a powerful

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service