Skip to Content
Merck
All Photos(3)

Key Documents

17-681

Sigma-Aldrich

ChIPAb+ Dimethyl-Histone H3 (Lys9) - ChIP Validated Antibody and Primer Set

clone CMA307, from mouse

Synonym(s):

H3K9me2, Histone H3 (di methyl K9)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.52
clone:
CMA307, monoclonal
application:
ChIP
WB
species reactivity:
human, vertebrates
technique(s):
ChIP: suitable
western blot: suitable
citations:
14

biological source

mouse

Quality Level

clone

CMA307, monoclonal

species reactivity

human, vertebrates

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Dimethyl-Histone H3 (Lys9) set includes the Anti-dimethyl-Histone H3 (Lys9) antibody, a negative control antibody (purified Mouse IgG), and qPCR primers which amplify a 110 bp region within the promoter of the human β-globin gene. The dimethyl-histone H3 (Lys9) and negative control antibodies are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of dimethyl-histone H3 (Lys9) associated chromatin.

Specificity

Recognizes histone H3, Mr 17 kDa, dimethylated at lysine 9.
This peptide sequence is identical in a wide range of animal and plant species.

Immunogen

Epitope: a.a. 1-18
The dimethyl-histone H3 (Lys9) purified antibody is made against a synthetic peptide (dimethylated at Lys9) corresponding to amino acids 1-18 of histone H3.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1 X 106 cell equivalents per IP) was subjected to chromatin immunoprecipitation using 2 µg of either a normal mouse IgG or Anti-dimethyl-Histone H3 (Lys9) antibody and the Magna ChIP G (Cat. #17-611) Kit. Successful immunoprecipitation of dimethyl-histone H3 (Lys9) associated DNA fragments was verified by qPCR using β-globin ChIP Primers versus Control Primers (Please see figures). Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna G ChIP (Cat. #17-408) or EZ-ChIP (Cat. #17-371) protocol for experimental details.

Dot Blot Analysis: Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665), which contain histone peptides with various modifications were probed with Anti-dimethyl H3 (Lys9) at 2.0ug/mL (1:500) dilution. Proteins were visualized using a Donkey anti-mouse IgG conjugated to HRP and a chemiluminescence detection system.

Western Blot Analysis:
Recombinant Histone H3 (Lane 1) and HeLa acid extract (Lane 2) were resolved by electrophoresis, transferred to PVDF membrane and probed with Anti-dimethyl Histone H3 (Lys9), clone CMA307 (0.1 μg/ml). Proteins were visualized using a goat anti-mouse secondary antibody conjugated to HRP and a chemiluminescence detection system.

Multiplexing:
This antibody specifically recognizes histone H3 dimethylated on Lys9 by Luminex assay.

ELISA: This antibody has been shown by an outside laboratory to be suitable for ELISA.

Immunocytochemistry: This antibody has been shown by an outside laboratory to be suitable for immunocytochemistry.

Immunoprecipitation: This antibody has been shown by an outside laboratory to be suitable for immunoprecipitation.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This ChIPAb+ Dimethyl-Histone H3 (Lys9) -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

25 assays per kit, ~2μg per chromatin immunoprecipitation

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1 X 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 2 µg of either a normal mouse IgG or Anti-dimethyl-Histone H3 (Lys9) antibody and the Magna ChIP G (Cat. #17-611) Kit. Successful immunoprecipitation of dimethyl-histone H3 (Lys9) associated DNA fragments was verified by qPCR using ChIP Primers B-Globin (Please see figures).
Please refer to the EZ-Magna G ChIP (Cat. #17-409) or EZ-ChIP (Cat. #17-371) protocol for experimental details.

Target description

Dimethyl-histone H3 at ~17 kDa

Physical form

Anti-dimethyl-Histone H3 (Lys9) (mouse monoclonal IgG1қ, Clone CMA307). One vial containing 50 μg of protein G purified antibody in 50 μL PBS containing 0.05% sodium. Store at -20°C.

Normal Mouse IgG. Two vials containing 25 μg purified Mouse IgG in 25 μL storage buffer containing 0.1% sodium azide. Store at -20°C.

ChIP Primers B-Globin. One vial containing 75 μL of 5 μM of each primer specific for human β-globin.
Store at -20°C.
FOR: AGG ACA GGT ACG GCT GTC ATC
REV: TTT ATG CCC AGC CCT GGC TC
Format: Purified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Aliquot upon initial thaw, avoid freeze thaw cycles.

Analysis Note

Control
Included negative control mouse IgG antibody and control primers specific for human β-globin promoter.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emanuela Sani et al.
Genome biology, 14(6), R59-R59 (2013-06-19)
In arid and semi-arid environments, drought and soil salinity usually occur at the beginning and end of a plant's life cycle, offering a natural opportunity for the priming of young plants to enhance stress tolerance in mature plants. Chromatin marks
Marcela A Bennesch et al.
Nucleic acids research, 44(18), 8655-8670 (2016-06-22)
The estrogen receptor α (ERα) is a transcription factor that can be directly activated by estrogen or indirectly by other signaling pathways. We previously reported that activation of the unliganded ERα by cAMP is mediated by phosphorylation of the transcriptional
Jin Rong Ow et al.
Scientific reports, 6, 34163-34163 (2016-09-27)
In this study, we demonstrate that the lysine methyltransferase G9a inhibits sarcomere organization through regulation of the MEF2C-HDAC5 regulatory axis. Sarcomeres are essential for muscle contractile function. Presently, skeletal muscle disease and dysfunction at the sarcomere level has been associated
Huacheng Luo et al.
Cancer cell, 36(6), 645-659 (2019-12-02)
Long non-coding RNAs (lncRNAs) are critical for regulating HOX genes, aberration of which is a dominant mechanism for leukemic transformation. How HOX gene-associated lncRNAs regulate hematopoietic stem cell (HSC) function and contribute to leukemogenesis remains elusive. We found that HOTTIP
Alba Fernández-Sánchez et al.
Epigenetics, 8(1), 66-78 (2012-12-14)
The human activating receptor NKG2D is mainly expressed by NK, NKT, γδ T and CD8(+) T cells and, under certain conditions, by CD4(+) T cells. This receptor recognizes a diverse family of ligands (MICA, MICB and ULBPs 1-6) leading to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service