Skip to Content
Merck
All Photos(3)

Documents

17-10147

Sigma-Aldrich

LentiBrite GFP-HMGB1 Lentiviral Biosensor

Synonym(s):

High mobility group protein B1, High mobility group protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
34360190
NACRES:
NA.51

manufacturer/tradename

Chemicon®
LentiBrite

Quality Level

technique(s)

cell based assay: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
transfection: suitable

UniProt accession no.

detection method

fluorometric

shipped in

dry ice

Gene Information

human ... HMGB1(3146)

General description

Read our application note in Nature Methods!
http://www.nature.com/app_notes/nmeth/2012/121007/pdf/an8620.pdf
(Click Here!)

Learn more about the advantages of our LentiBrite Lentiviral Biosensors! Click Here

Biosensors can be used to detect the presence/absence of a particular protein as well as the subcellular location of that protein within the live state of a cell. Fluorescent tags are often desired as a means to visualize the protein of interest within a cell by either fluorescent microscopy or time-lapse video capture. Visualizing live cells without disruption allows researchers to observe cellular conditions in real time.

Lentiviral vector systems are a popular research tool used to introduce gene products into cells. Lentiviral transfection has advantages over non-viral methods such as chemical-based transfection including higher-efficiency transfection of dividing and non-dividing cells, long-term stable expression of the transgene, and low immunogenicity.

EMD Millipore is introducing LentiBrite Lentiviral Biosensors, a new suite of pre-packaged lentiviral particles encoding important and foundational proteins of autophagy, apoptosis, and cell structure for visualization under different cell/disease states in live cell and in vitro analysis.
  • Pre-packaged, fluorescently-tagged with GFP & RFP
  • Higher efficiency transfection as compared to traditional chemical-based and other non-viral-based transfection methods
  • Ability to transfect dividing, non-dividing, and difficult-to-transfect cell types, such as primary cells or stem cells
  • Non-disruptive towards cellular function

EMD Millipore’s LentiBrite GFP-HMGB1 lentiviral particles provide bright fluorescence and precise localization to enable live cell analysis of HMGB1 translocation in difficult-to-transfect cell types.
HMBG1 (high mobility box group 1) functions in the nucleus of most cells to stabilize nucleosomes and enhance binding of transcription factor complexes to DNA. In certain activated inflammatory cells, HMGB1 is actively secreted, whereas necrotic cells passively release HMGB1. In both cases, extracellular HMGB1 can bind to multiple immune cell types via RAGE (receptor for advanced glycation end products) to stimulate chemotaxis and cytokine production, which, depending on the context, results in inflammation, wound healing, or immunogenic tumor cell death. Cells undergoing autophagy also display translocation of HMGB1, from the nucleus to the cytoplasm. Expression of genetic fusions between HMGB1 and fluorescent proteins permits monitoring of HMGB1 translocation in real time.
EMD Millipore’s LentiBrite GFP-HMGB1 lentiviral particles provide bright fluorescence and precise localization to enable live cell analysis of HMGB1 translocation in difficult-to-transfect cell types.

Application

Fluorescence Microscopy Imaging:
(See Figure 1 in datasheet)
U2OS cells were plated in a chamber slide and transduced with lentiviral particles at an MOI of 10 for 24 hours. After media replacement and 48 hours further incubation, cells were fixed with formaldehyde and mounted. Images were obtained by oil immersion wide-field fluorescence microscopy. The GFP-HMGB1 displays in the nucleus of cells.

Immunocytochemistry Comparison and Inhibitor Analysis:
(See Figure 2 in datasheet)
Similar to Figure 1 (See datasheet), HeLa cells were plated in a chamber slide and transduced with lentiviral particles at an MOI of 10 for 24 hours. After 24 hours, media was replaced and cells were then further incubated for 48 hours. Immunocytochemical staining (blue) of the same fields of view with a nuclear stain (DAPI) reveals similar expression patterns to the protein-GFP (green).

Difficult-to-transfect Cell Types:
(See Figure 3 in datasheet)
Primary cell types HUVEC and Human mesenchymal stem cells (HuMSC) were plated in chamber slides and transduced with lentiviral particles at an MOI of 20 for 24 hours.

For optimal fluorescent visualization, it is recommended to analyze the target expression level within 24-48 hrs after transfection/infection for optimal live cell analysis, as fluorescent intensity may dim over time, especially in difficult-to-transfect cell lines. Infected cells may be frozen down after successful transfection/infection and thawed in culture to retain positive fluorescent expression beyond 24-48 hrs. Length and intensity of fluorescent expression varies between cell lines. Higher MOIs may be required for difficult-to-transfect cell lines.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Components

TagGFP2-HMGB Lentivirus:
One vial containing 25 µL of lentiviral particles at a minimum of 3 x 10E8 infectious units (IFU) per mL.
For lot-specific titer information, please see lot specific “Viral Titer” in the product specifications of the datasheet.


Promoter
EF-1 (Elongation Factor-1)


Multiplicty of Infection (MOI)
MOI = Ratio of # of infectious lentiviral particles (IFU) to # of cells being infected.
Typical MOI values for high transduction efficiency and signal intensity are in the range of 20-40. For this target, some cell types may require lower MOIs (e.g., HT-1080, HeLa, U2OS), while others may require higher MOIs (e.g., human umbilical vein endothelial cells (HUVEC), human mesenchymal stem cells (HuMSC)).
NOTE: MOI should be titrated and optimized by the end user for each cell type and lentiviral target to achieve desired transduction efficiency and signal intensity.

Quality

Evaluated by transduction of HT-1080 cells and fluorescent imaging performed for assessment of target localization and transduction efficiency.

Physical form

PEG precipitation

Storage and Stability

Storage and Handling
Lentivirus is stable for at least 4 months from date of receipt when stored at -80°C. After first thaw, place immediately on ice and freeze in working aliquots at -80°C. Frozen aliquots may be stored for at least 2 months. Further freeze/thaws may result in decreased virus titer and transduction efficiency.

IMPORTANT SAFETY NOTE
Replication-defective lentiviral vectors, such as the 3rd Generation vector provided in this product, are not known to cause any diseases in humans or animals. However, lentiviruses can integrate into the host cell genome and thus pose some risk of insertional mutagenesis. Material is a Risk Group 2 and should be handled under BSL2 controls. A detailed discussion of biosafety of lentiviral vectors is provided in Pauwels, K. et al. (2009). State-of-the-art lentiviral vectors for research use: Risk assessment and biosafety recommendations. Curr. Gene Ther. 9: 459-474.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Fluorescent lentiviral particles encoding important GFP/RFP fusion proteins related to autophagy, apoptosis, and cell structure that enables live cell imaging.

Fluorescent lentiviral particles encoding important GFP/RFP fusion proteins related to autophagy, apoptosis, and cell structure that enables live cell imaging.

Fluorescent lentiviral particles encoding important GFP/RFP fusion proteins related to autophagy, apoptosis, and cell structure that enables live cell imaging.

Fluorescent lentiviral particles encoding important GFP/RFP fusion proteins related to autophagy, apoptosis, and cell structure that enables live cell imaging.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service