Skip to Content
Merck
All Photos(2)

Key Documents

1.46634

Millipore

Buffered NaCl Peptone Solution + 0.1% Tween® 80

bottle of, ready-to-use, bottle volume 450 mL , filling volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type screw cap with 3 septa
ready-to-use

packaging

bottle of

technique(s)

direct inoculation: suitable
microbiological culture: suitable

bottle capacity

500 mL

bottle volume

450 mL , filling volume

pH

7.0±0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

components

tween (0.1%)

storage temp.

2-25°C

suitability

nonselective for

General description

Buffered NaCl Peptone Solution with 0.1% Tween® 80 is a low peptone content buffer supplemented with phosphate buffer and sodium chloride. The unique formula maintains viability of test microorganisms present in test samples.
Polysorbate 80 (Tween® 80), is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines, dequadin, benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols and PHB esters.

Application

Buffered NaCl Peptone Solution with 0.1% Tween® 80 is used for the microbial examination of non-sterile samples. It is suitable for dilution, sample preparation and sample washing.

Features and Benefits

  • Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements.
  • Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy, and significantly reducing the risk of incorrect interpretation and false results.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service