Skip to Content
Merck
All Photos(2)

Key Documents

1.46386

Millipore

Fluid Thioglycollate Medium

ready-to-use, bottle volume 100 mL , filling volume

Synonym(s):

Fluid Thioglycollate Medium acc EP+USP, Fluid Thioglycollate Medium, FTM, Thioglycollate Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile

form

liquid (ready-to-use)

shelf life

12 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

bottle of
single packed of (100 ml in 125 ml bottle with red screw cap and septum (10 bottles per box))

technique(s)

sterile filtration: suitable

bottle capacity

125 mL

bottle volume

100 mL , filling volume

pH

7.1

application(s)

cosmetics
food and beverages
microbiology
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

aerobic bacteria
anaerobic bacteria

Looking for similar products? Visit Product Comparison Guide

General description

Fluid Thioglycolate Medium is a universal complex medium with thioglycollate and L-Cystine to reduce the redox potential of the culture medium and create an anaerobic atmosphere. They also inactivate mercury and other heavy metal compounds. Agar in the medium further reduces the rapid diffusion of oxygen but may lead to slight turbidity in larger volumes (filled bottles). Resazurin indicates the reduction potential of the medium. An increased concentration of oxygen is indicated by a color change to pink.

Application

Fluid Thioglycolate Medium is suitable for the isolation and cultivation of fastidious anaerobic as well as for aerobic microorganisms. It is used for sterility control of pharmaceutical products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service