14-329-M
JNK2α2/SAPK1a Protein, active, 10 µg
Active, N-terminal His-tagged, full length human JNK2α2/SAPK1a, for use in Kinase Assays.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41
biological source
human
Quality Level
mol wt
Mw 49.2 kDa
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
NCBI accession no.
UniProt accession no.
Gene Information
human ... MAPK9(5601)
General description
N-terminal His-tagged, full length human JNK2α2/SAPK1a
Product Source: Expressed in Sf21 cells
Biochem/physiol Actions
Protein Target: JNK2α2/SAPK1a
Target Sub-Family: CMGC
Packaging
Also available in 250 µg size --call for pricing and availability and reference catalog number 14-329M when ordering the 250 µg size.
Quality
Routinely evaluated by phosphorylation of ATF2 (amino acids 19-96)
Physical form
Ni2+/NTA-agarose
Storage and Stability
6 months at -70°C
Other Notes
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Girish V Putcha et al.
Neuron, 38(6), 899-914 (2003-06-24)
Trophic factor deprivation (TFD) activates c-Jun N-terminal kinases (JNKs), culminating in coordinate AP1-dependent transactivation of the BH3-only BCL-2 proteins BIM(EL) and HRK, which in turn are critical for BAX-dependent cytochrome c release, caspase activation, and apoptosis. Here, we report that
Y Zhang et al.
The Journal of biological chemistry, 276(24), 20913-20923 (2001-03-30)
Ultraviolet light A (UVA) plays an important role in the etiology of human skin cancer, and UVA-induced signal transduction has a critical role in UVA-induced skin carcinogenesis. The upstream signaling pathways leading to p70(S6K) phosphorylation and activation are not well
S P Zhong et al.
The Journal of biological chemistry, 275(28), 20980-20984 (2000-05-12)
Histone H3 is the core protein of the nucleosome. Phosphorylation of H3 involves immediate early gene expression, chromatin remodeling, and chromosome condensation during mitosis. Very recently, Rsk2 or MSK1 kinase-mediated phosphorylation of H3 at serine 10 was reported. In the
S Gupta et al.
The EMBO journal, 15(11), 2760-2770 (1996-06-03)
The JNK protein kinase is a member of the MAP kinase group that is activated in response to dual phosphorylation on threonine and tyrosine. Ten JNK isoforms were identified in human brain by molecular cloning. These protein kinases correspond to
S Gupta et al.
Science (New York, N.Y.), 267(5196), 389-393 (1995-01-20)
Treatment of cells with pro-inflammatory cytokines or ultraviolet radiation causes activation of the c-Jun NH2-terminal protein kinase (JNK). Activating transcription factor-2 (ATF2) was found to be a target of the JNK signal transduction pathway. ATF2 was phosphorylated by JNK on
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service