Skip to Content
Merck
All Photos(2)

Key Documents

14-276-M

Sigma-Aldrich

Akt1/PKBα Protein, active, 15 µg

Active, recombinant full-length human Akt1, containing N-terminal His6 tag, for use in IP-Kinase Assays & Kinase Assays.

Synonym(s):

Protein kinase B alpha, Proto-oncogene c-Akt, RAC, RAC-PK-alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.26

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

form

liquid

shelf life

1 yr

mol wt

Mw 59.9 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (IP-kinase)
activity assay: suitable (kinase)

solubility

soluble

UniProt accession no.

storage temp.

−70°C

Gene Information

human ... AKT1(207)

General description

Research area: CELL SIGNALING

Product Source: Expressed by baculovirus in Sf21 insect cells

Recombinant full-length human Akt1, containing N-terminal His6-tag.

Biochem/physiol Actions

Protein Target: Akt1/PKBα

Target Sub-Family: AGC

Akt, also known as protein Kinase B (PKB), is a crucial protein involved in various cell signaling pathways. It modifies and controls the activities of numerous proteins involved in a wide range of cellular and physiological processes. Some of these processes include cell survival, apoptosis, cell proliferation, transcription, cell migration, and metabolism. Akt also helps in influencing angiogenesis, and normal development of embryos. Akt regulates cell survival through the phosphorylation of mitogen-activated protein kinase kinase kinase 5 (MAP3K5), which is involved in the cellular apoptotic response. The Akt occupies a pivotal position in the phosphoinositide 3-kinase-protein kinase B-mammalian target of rapamycin (PI3K-PKB-mTOR) signaling pathway. Genetic mutations occurring upstream of Akt in this cascade can result in uncontrolled cell proliferation and the development of cancer. In addition abberent Akt activation is associated with the development of type-2 diabetes.

Packaging

Also available in 250μg size, please inquire for pricing and availability.

Quality

Routinely evaluated by phosphorylation of Crosstide. Please refer to the Certificate of Analysis for more information.

Physical form

Ni2+/NTA and glutathione agarose
Supplied in 50mM Tris/HCl pH7.5, 150mM NaCl, 150mM imidazole, 1mM benzamidine, 0.2mM PMSF, 0.1mM EGTA, 0.07% 2-mercaptoethanol, 0.02% Brij 35, 270mM sucrose. Frozen solution.

Storage and Stability

1 year at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin Receptor Signaling
Laychock SG
Encyclopedia of Hormones, 368-380 (2003)
AKT/PKB signaling: navigating downstream
Manning BD, et al.
Cell, 129(7), 1261-1274 (2007)
Structural Biology and Anticancer Drug Design
Pathuri P, et al.
Journal of Separation Science, 2, 121-141 (2014)
Dosage-dependent effects of Akt1/protein kinase Balpha (PKBalpha) and Akt3/PKBgamma on thymus, skin, and cardiovascular and nervous system development in mice
Yang Z-Z, et al.
Molecular and Cellular Biology, 25(23), 10407-10418 (2005)
Madhavi J Rane et al.
The Journal of biological chemistry, 278(30), 27828-27835 (2003-05-13)
Activation of the serine-threonine kinase Akt by cytokines, chemokines, and bacterial products delays constitutive neutrophil apoptosis, resulting in a prolonged inflammatory response. We showed previously that Akt exists in a signaling complex with p38 MAPK, MAPK-activated protein kinase-2 (MAPKAPK-2), and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service