Skip to Content
Merck
All Photos(6)

Key Documents

1.09728

Supelco

Pyridine

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Pyridine, Azine, Azabenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
MDL number:
UNSPSC Code:
12191501
EC Index Number:
203-809-9
NACRES:
NB.21
grade:
ACS reagent
for analytical purposes
Assay:
≥99.5% (GC)
bp:
115 °C/1013 hPa
vapor pressure:
26.7 hPa ( 25 °C)
Pricing and availability is not currently available.

grade

ACS reagent
for analytical purposes

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

26.7 hPa ( 25 °C)

product line

EMSURE®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

482 °C

potency

891 mg/kg LD50, oral (Rat)
1121 mg/kg LD50, skin (Rabbit)

impurities

≤0.002% Ammonia (NH3)
≤0.01% Piperidine (GC)
≤0.1% Water
≤0.2% 2-Picoline (GC)

evapn. residue

≤0.002%

color

clearAPHA: ≤10

pH

8.81 (20 °C in H2O)

bp

115 °C/1013 hPa

mp

-42 °C

transition temp

flash point 17 °C

density

0.982 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.0005%
sulfate (SO42-): ≤0.0005%

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

storage temp.

2-30°C

SMILES string

n1ccccc1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application


  • Enantioselective separation of amino acids as biomarkers indicating life in extraterrestrial environments.: Research on using pyridine-based chromatography for the enantioselective separation of amino acids, which can help in the search for extraterrestrial life by analyzing amino acid chirality (Pietrogrande, 2013).

Linkage

Discover EMSURE - to be more than sure!

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Solubility in water: conforms
Boiling point: 114 - 116 °C
Reducing substances: conforms
Ammonia (NH₃): ≤ 0.002 %
2-Picoline (GC): ≤ 0.2 %
Piperidine (GC): ≤ 0.01 %
Chloride (Cl): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.0005 %
Al (Aluminium): ≤ 0.00005 %
B (Boron): ≤ 0.000002 %
Ba (Barium): ≤ 0.00001 %
Ca (Calcium): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.000005 %
Co (Cobalt): ≤ 0.000002 %
Cr (Chromium): ≤ 0.000002 %
Cu (Copper): ≤ 0.000002 %
Fe (Iron): ≤ 0.00001 %
Mg (Magnesium): ≤ 0.00001 %
Mn (Manganese): ≤ 0.000002 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.00001 %
Sn (Tin): ≤ 0.00001 %
Zn (Zinc): ≤ 0.00001 %
Evaporation residue: ≤ 0.002 %
Water: ≤ 0.1 %

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service