Skip to Content
Merck
All Photos(1)

Key Documents

1.09217

Sigma-Aldrich

Gram′s safranine solution

for the Gram staining method

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

form

liquid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

transition temp

flash point 49 °C

density

0.98 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

This ready-to-use Gram′s safranine solution-for the Gram staining method, is used in bacteriology. The Gram staining allows a fast differentiation of bacteria in Gram-positive and Gram-negative. The mureine structure of the bacteria walls is the basis of the color affinity. Bacteria will be stained with Gram′s crystal violet solution (Product number 1.09218) - an aniline dye - in the first step. After the treatment with iodinesolution (Lugol′s solution stabilized with PVP, Product number 1.00567), a dye-iodine complex will form. During the decolorizing step (Gram′s decolorizing solution, 1.10218), this complex stays in the multilayer mureine structures of the Gram-positive bacteria and they will appear blue-violet. Gram-negative bacteria have a monolayer mureine structure only, the dye-iod complex does not stay bound to the cellwall, they will be decolorized . Gram-negative bacteria will be counterstained by Gram′s safranin solution and appear pink to red.
A bottle with 500 ml Gram′s safranin solution is sufficient for staining up to 250 slides, it is also available as part of the Gram-color stain set, (Product number 1.11885). It is an IVD product and CE registered. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F

Flash Point(C)

49 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. I have Gram's safranin solution for microscopy MERCK want to know about the conc. Of this product.

    1 answer
    1. The concentration of the Gram's Safranine solution is considered proprietary since the concentration is not listed in either the User's Guide or the Safety Data Sheet. Most textbook formulations for Grams Safranin will list the concentration as somewhere between 0.25% to 0.6% dye content.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service