All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
EC Index Number:
231-175-3
NACRES:
NA.21
form:
sticks
Recommended Products
Assay
≥99.9% (complexometric)
Quality Level
form
sticks
cation traces
As: ≤0.00001%
Cd: ≤0.005%
Cu: ≤0.002%
Fe: ≤0.005%
Pb: ≤0.005%
Sn: ≤0.001%
storage temp.
2-30°C
SMILES string
[Zn]
InChI
1S/Zn
InChI key
HCHKCACWOHOZIP-UHFFFAOYSA-N
General description
sticks, triangular cross section about 8 mm for analysis EMSURE®
Analysis Note
Assay (complexometric): ≥ 99.9 %
As (Arsenic): ≤ 0.00001 %
Cd (Cadmium): ≤ 0.005 %
Cu (Copper): ≤ 0.002 %
Fe (Iron): ≤ 0.005 %
Pb (Lead): ≤ 0.005 %
Sn (Tin): ≤ 0.001 %
As (Arsenic): ≤ 0.00001 %
Cd (Cadmium): ≤ 0.005 %
Cu (Copper): ≤ 0.002 %
Fe (Iron): ≤ 0.005 %
Pb (Lead): ≤ 0.005 %
Sn (Tin): ≤ 0.001 %
Legal Information
EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service