Skip to Content
Merck
All Photos(1)

Key Documents

1.08087

Supelco

Potassium sodium tartrate tetrahydrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Potassium sodium tartrate tetrahydrate, Sodium potassium tartrate, Tartaric acid potassium sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4KNaO6 · 4H2O
CAS Number:
Molecular Weight:
282.22
MDL number:
UNSPSC Code:
12352107
EC Index Number:
206-156-8
NACRES:
NA.21
grade:
ACS reagent
form:
solid

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.
suitable for SM 4500 - NH3

product line

EMSURE®

Assay

99.0-102.0% (alkalimetric)

form

solid

impurities

≤0.005% Insoluble matter

pH

7.0-8.5 (25 °C, 50 g/L in H2O)

mp

70-80 °C

bulk density

1000 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.004%
Cu: ≤0.0005%
Fe: ≤0.0005%
NH4+: ≤0.002%
Pb: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[K+].[Na+].[O-]C(=O)[C@H](O)[C@@H](O)C(=O)[O-].O.O.O.O

InChI

1S/C4H6O6.K.Na.4H2O/c5-1(3(7)8)2(6)4(9)10;;;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;4*1H2/q;2*+1;;;;/p-2/t1-,2-;;;;;;/m1....../s1

InChI key

VZOPRCCTKLAGPN-ZFJVMAEJSA-L

Looking for similar products? Visit Product Comparison Guide

Linkage

Replaces: PX1590; PX1590-1; PX1590-2; PX1590-5; PX1590-11;

Analysis Note

Assay (alkalimetric): 99.0 - 102.0 %
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water; 25 °C): 7.0 - 8.5
Chloride (Cl): ≤ 0.0005 %
Phosphate (PO₄): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.004 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
NH₄ (Ammonium): ≤ 0.002 %
Pb (Lead): ≤ 0.0005 %
Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service