Skip to Content
Merck
All Photos(1)

Documents

1.05413

Millipore

ROGOSA agar

Lactobacillus selective agar for microbiology

Synonym(s):

Selective agar for lactobacillus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

solubility

74.5 g/L

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

Lactobacillus spp.

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish-brown
Recovery on test medium (Lactobacillus acidophilus ATCC 4356 (WDCM 00098)): ≥ 70 %
Recovery on test medium (Lactobacillus casei ATCC 393 (WDCM 00100)): ≥ 70 %
Recovery on test medium (Lactobacillus fermentum ATCC 9338): ≥ 70 %
Recovery on test medium (Lactobacillus plantarum ATCC 8014): ≥ 70 %
Recovery on test medium (Bifidobacterium bifidum ATCC 11863): ≥ 70 %
Recovery on test medium (Escherichia coli ATCC 11775 (WDCM 00090)): ≤ 0.01 %
Recovery on test medium (Proteus hauseri ATCC 13315): ≤ 0.01 %
Recovery on test medium (Enterococcus faecalis ATCC 11700): ≤ 0.01 %
Incubation: 48-72 hrs.; 35 °C; aerobic (Bifidobacterium anaerobic)

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service