Skip to Content
Merck
All Photos(1)

Key Documents

1.05067

Supelco

Potassium nitrite

cryst. for analysis EMSURE® ACS

Synonym(s):

Potassium nitrite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO2
CAS Number:
Molecular Weight:
85.10
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-832-4
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
2810 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

≥97.0% (cerimetric)

form

solid

impurities

≤0.01% Insoluble matter

pH

7-10 (20 °C, 50 g/L in H2O)

mp

407 °C

solubility

2810 g/L

density

1.92 g/cm3 at 20 °C

bulk density

980 kg/m3

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.003%
Fe: ≤0.0005%
Mg: ≤0.002%
Na: ≤0.5%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[K+].N(=O)[O-]

InChI

1S/K.HNO2/c;2-1-3/h;(H,2,3)/q+1;/p-1

InChI key

BXNHTSHTPBPRFX-UHFFFAOYSA-M

Application

  • Suitability evaluation of new endogenous biomarkers for the identification of nitrite-based urine adulteration in mass spectrometry methods.: This paper assesses novel biomarkers for spotting urine tampering using potassium nitrite, critical for maintaining integrity in analytical testing scenarios (Steuer et al., 2019).
  • 2-Nitro-6-monoacetylmorphine: potential marker for monitoring the presence of 6-monoacetylmorphine in urine adulterated with potassium nitrite.: Introduces a potential marker for detecting adulteration in urine tests, essential for chemists focused on drug testing accuracy (Luong et al., 2012).

Analysis Note

Assay (cerimetric): ≥ 97.0 %
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water): 7.0 - 10.0
Chloride (Cl): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.003 %
Fe (Iron): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.002 %
Na (Sodium): ≤ 0.5 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service