Skip to Content
Merck
All Photos(1)

Key Documents

1.03420

Sigma-Aldrich

Chloroform-D1

deuteration degree min. 99.8% for NMR spectroscopy (stabilized with silver) MagniSolv

Synonym(s):

Chloroform-D1, Deuterochloroform

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCl3D
CAS Number:
Molecular Weight:
120.38
MDL number:
UNSPSC Code:
12142201
EC Index Number:
212-742-4
NACRES:
NA.22
grade:
NMR grade
technique(s):
NMR: suitable
bp:
60 °C/1013 hPa
vapor pressure:
211 hPa ( 20 °C)
Pricing and availability is not currently available.

grade

NMR grade

Quality Level

vapor pressure

211 hPa ( 20 °C)

form

liquid

potency

695 mg/kg LD50, oral (Rat)
>3980 mg/kg LD50, skin (Rabbit)

technique(s)

NMR: suitable

bp

60 °C/1013 hPa

mp

-64.1 °C

density

1.50 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)[2H]

InChI

1S/CHCl3/c2-1(3)4/h1H/i1D

InChI key

HEDRZPFGACZZDS-MICDWDOJSA-N

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service