1.02888
Cyclohexanone
EMPLURA®
Synonym(s):
Cyclohexanone, Pimelic ketone
About This Item
vapor pressure
4.5 hPa ( 20 °C)
Quality Level
product line
EMPLURA®
Assay
≥99% (GC)
form
liquid
autoignition temp.
430 °C
expl. lim.
1.1 % (v/v)
dilution
(low-cost alternative, for basic applications)
impurities
≤0.2% Water
pH
7 (20 °C, 70 g/L in H2O)
bp
155 °C/1013 hPa
mp
-31 °C
transition temp
flash point 43 °C
solubility
90 g/L
density
0.95 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O=C1CCCCC1
InChI
1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2
InChI key
JHIVVAPYMSGYDF-UHFFFAOYSA-N
Related Categories
General description
Analysis Note
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 0.946 - 0.948
Water: ≤ 0.2 %
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
111.2 °F - closed cup
Flash Point(C)
44 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service