1.01774
Petroleum benzine boiling range 60-80°C
for analysis EMSURE®
Synonym(s):
Petroleum ether, Petroleum spirit, Petroleum ether
About This Item
Recommended Products
grade
reagent grade
Quality Level
vapor pressure
200 hPa ( 20 °C)
product line
EMSURE®
form
liquid
autoignition temp.
240 °C
expl. lim.
1.0-7.4 % (v/v)
dilution
(for analytical testing)
impurities
≤0.000002% B (Boron)
≤0.000002% Co (Cobalt)
≤0.000002% Cr (Chromium)
≤0.000005% Cd (Cadmium)
≤0.00001% Ba (Barium)
≤0.00005% Al (Aluminium)
≤0.00005% Ca (Calcium)
≤0.0003 meq/g Acidity
≤0.005% Aromatics (as benzene)
≤0.005% Sulfur compounds (as S)
≤0.01% Water
evapn. residue
≤0.001%
bp
60-80 °C/1013 hPa
iodine value
≤0.3
transition temp
flash point -26 °C
density
0.68 g/cm3 at 20 °C
cation traces
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%
storage temp.
2-30°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Linkage
Other Notes
Legal Information
accessory
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation
Target Organs
Nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-14.8 °F
Flash Point(C)
-26 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service