Skip to Content
Merck
All Photos(1)

Key Documents

1.01261

Supelco

Aniline

for analysis EMSURE®

Synonym(s):

Aniline, Aminobenzene, Phenylamine, Benzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13
MDL number:
UNSPSC Code:
12352116
EC Index Number:
200-539-3
NACRES:
NA.21
Assay:
≥99.5% (GC)
bp:
184 °C/1013 hPa
vapor pressure:
0.5 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

0.5 hPa ( 20 °C)

Quality Level

product line

EMSURE®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

540 °C

potency

840 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-11 % (v/v)

impurities

≤0.003% Nitrobenzene (GC)
≤0.005% Sulfated ash
≤0.1% Water

pH

8.8 (20 °C, 36 g/L in H2O)

bp

184 °C/1013 hPa

mp

-6.2 °C

transition temp

flash point 76 °C

solubility

36 g/L

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Nc1ccccc1

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2

InChI key

PAYRUJLWNCNPSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Protocols

Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.

Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.

Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.

Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service