1.01203
Potassium chloride solution
nominal 1.41 mS/cm, certified reference material for the measurement of electrolytic conductivity (c=0.01 mol/l) Certipur®
Synonym(s):
Potassium chloride solution
About This Item
Recommended Products
Product Name
Potassium chloride solution (nominal 1.41 mS/cm), nominal 1.41 mS/cm, certified reference material for the measurement of electrolytic conductivity (c=0.01 mol/l) Certipur®
grade
certified reference material
Quality Level
Agency
ISO (17025, 17034)
product line
Certipur®
pH
7 (20 °C in H2O)
bp
100 °C/1013 hPa
application(s)
environmental
format
single component solution
storage temp.
15-25°C
SMILES string
[Cl-].[K+]
InChI
1S/ClH.K/h1H;/q;+1/p-1
InChI key
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
Analysis Note
Legal Information
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service