Skip to Content
Merck
All Photos(5)

Key Documents

1.00979

Supelco

Isoamyl alcohol

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1718835
MDL number:
UNSPSC Code:
12191502
EC Index Number:
204-633-5
NACRES:
NB.21
grade:
ACS reagent
for analytical purposes
bp:
130 °C (lit.)
vapor pressure:
2 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent
for analytical purposes

Quality Level

Agency

reag. Ph. Eur.

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 20 °C)

product line

EMSURE®

form

liquid

autoignition temp.

644 °F

potency

>5000 mg/kg LD50, oral (Rat)
>3000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-9 %, 100 °F

dilution

(for analytical testing)

impurities

≤0.0001% Furfurol
≤0.005% Free acid (as CH3COOH)
≤0.05% Carbonyl compounds (as CO)
≤0.06% Acids,esters (as pentylacetate)
≤0.2% Water
≤0.5% n-Amyl alcohol (GC)
≥99% Total isomeric compounds (GC)

evapn. residue

≤0.002%

color

clearAPHA: ≤10

refractive index

n20/D 1.406 (lit.)

pH

7 (20 °C, 25 g/L in H2O)

kinematic viscosity

5.32 cSt(20 °C)

bp

130 °C (lit.)

mp

−117 °C (lit.)

transition temp

flash point 43.5 °C

solubility

25 g/L

density

0.809 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

SMILES string

CC(C)CCO

storage temp.

2-30°C

InChI

1S/C5H12O/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

PHTQWCKDNZKARW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

110.3 °F - closed cup

Flash Point(C)

43.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service