1.00397
N,N-Dimethylformamide
for peptide synthesis
Synonym(s):
DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide, DMF, NSC 5356
About This Item
vapor density
2.5 (vs air)
Quality Level
vapor pressure
2.7 mmHg ( 20 °C)
Assay
≥99.9% (GC)
form
liquid
autoignition temp.
833 °F
potency
3010 mg/kg LD50, oral (Rat)
1500 mg/kg LD50, skin (Rabbit)
expl. lim.
15.2 %
dilution
(for synthesis)
impurities
≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤10 ppm Amines (Dimethylamine)
≤300 ppm Water
color
APHA: ≤10
refractive index
n20/D 1.430 (lit.)
pH
7 (20 °C, 200 g/L in H2O)
bp
153 °C (lit.)
mp
−61 °C (lit.)
transition temp
flash point 57.5 °C
solubility
soluble 1000 g/L
density
0.944 g/mL (lit.)
application(s)
peptide synthesis
storage temp.
2-30°C
SMILES string
[H]C(=O)N(C)C
InChI
1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3
InChI key
ZMXDDKWLCZADIW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Analysis Note
Identity (IR): conforms
Color: ≤ 10 Hazen
Acidity: ≤ 0.0005 meq/g
Alkalinity: ≤ 0.0002 meq/g
Amines (Dimethylamine): ≤ 10 ppm
Evaporation residue: ≤ 10
Water: ≤ 300 ppm
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
135.5 °F - closed cup
Flash Point(C)
57.5 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service