Skip to Content
Merck
All Photos(2)

Key Documents

1.00020

Supelco

Acetone

for liquid chromatography LiChrosolv®

Synonym(s):

Dimethyl ketone, Propanone, 2-Propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-662-2
NACRES:
NA.03
grade:
HPLC grade
technique(s):
HPLC: suitable
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)

grade

HPLC grade

Quality Level

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

description

isocratic

product line

LiChrosolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

465 °C (DIN 51794)

potency

5800 mg/kg LD50, oral (Rat)
20000 mg/kg LD50, skin (Rabbit)

expl. lim.

13.2 %

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.05% Water

evapn. residue

≤2.0 mg/L

transmittance

335 nm, ≥50%
340 nm, ≥80%
350 nm, ≥98%

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L in H2O)

capacity

1000  g/kg water absorbing capacity

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

transition temp

flash point <-20 °C

density

0.791 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

LiChrosolv® solvents show high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level. This makes them ideal for reproducible separations.

Application

Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .
Intended for HPLC and UHPLC-MS applications, either as an extraction solvent or as a mobile phase.
Reported for use in LC–MS/MS investigations of:
  • blood plasma and serum samples fractionated on several chromatography sorbents
  • peptides in animal biological samples

Features and Benefits

  • suitability tested and specified for UHPLC-MS and UHPLC-UV: for analytical flexibility
  • specified quality in positive and negative in positive and negative electrospray ionization (ESI) and atmospheric-pressure chemical ionization-mass spectrometry (APCI-MS) for lowest detection limits and confidence in analyses for all important MS modes (test 1)
—ESI/APCI (+) < 2 ppb
—ESI/APCI (-) < 10 ppb
  • lowest impurity profile: for interference-free baselines (test 2)
  • microfiltered through 0.2 μm filter (test 3) to provide:
—a long lifespan to filters and mechanical parts of HPLC systems
—reduced risk of column clogging

  • packed in borosilicate glass bottles for minimized metal ion contaminations
  • lowest levels of trace metal impurities for a minimum metal ion adduct formation
—<5 ppb
  • lowest level of polyethylene glycol (PEG) impurities in UHPLC-MS solvent lineup (PEG S/N signal-to-noise-ratio < 50)

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peptidome profiling dataset of ovarian cancer and non-cancer proximal fluids: Ascites and blood sera
Shender V, et al.
Data in Brief, 22, 557-562 (2019)
Proteomics-based identification of hypoxia-sensitive membrane-bound proteins in rat erythrocytes
Sidorenko S, et al.
Journal of Proteomics, 184, 25-33 (2018)
Peptidomics dataset: Blood plasma and serum samples of healthy donors fractionated on a set of chromatography sorbents
Arapidi G, et al.
Data in Brief, 18, 1204-1211 (2018)

Articles

This application note describes the HPTLC-based identification of fixed oils using octadecyl silyl silica gel as the coating substance, following USP 43 NF 38 monograph method containing USP general chapter <202>.

This application note describes the HPTLC-based identification of fixed oils using octadecyl silyl silica gel as the coating substance, following USP 43 NF 38 monograph method containing USP general chapter <202>.

This application note describes the HPTLC-based identification of fixed oils using octadecyl silyl silica gel as the coating substance, following USP 43 NF 38 monograph method containing USP general chapter <202>.

Simultaneous analysis of ascorbic acid and dehydroascorbic acid in various food samples by High Performance Thin Layer Chromatography (HPTLC).

See All

Protocols

Qualitative Thin Layer Chromatography Analysis of Flavonoids and Quantification of Terpene Lactones in Ginkgo Biloba Extracts and Tablets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service