Skip to Content
Merck
All Photos(2)

Key Documents

09-071

Sigma-Aldrich

Anti-G9a (BAT8) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

G9A histone methyltransferase, H3-K9-HMTase 3, HLA-B associated transcript 8, HLA-B-associated transcript 8, Histone H3-K9 methyltransferase 3, Lysine N-methyltransferase 1C, Protein G9a, ankyrin repeat-containing protein, chromosome 6 open reading frame

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human, mouse
technique(s):
western blot: suitable
citations:
10

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

canine, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EHMT2(10919)
mouse ... Ehmt2(110147)
rat ... Ehmt2(361798)

General description

G9a (BAT8 or Histone-lysine N-methyltransferase) is thought to be involved in intracellular protein-protein interaction and is a lysine-preferring histone methyltransferase. H3 Lys-9 methylation represents a specific tag for epigenetic transcriptional repression by recruiting HP1 proteins to methylated histones. The presence of the ankyrin repeats also suggests that G9a is involved in intracellular protein-protein interaction. Like Suv39h1, G9a transfers methyl groups to the lysine residues of histone H3, but with a 10- to 20-fold higher activity. G9a is localized to the nucleus but not in the repressive chromatin domains of centromeric loci, in which Suv39h1 family proteins were localized. This protein is probably targeted to histone H3 by different DNA-binding proteins like E2F6, MGA, MAX and/or DP1. G9a also methylates histone H1.

Specificity

This antibody recognizes G9a (BAT8).

Immunogen

KLH-conjugated linear peptide corresponding to G9a (BAT8).

Application

Anti-G9a (BAT8) Antibody is a rabbit polyclonal antibody for detection of G9a (BAT8) also known as G9A histone methyltransferase ,H3-K9-HMTase 3 ,HLA-B associated transcript 8 & has been validated in WB.
Western Blot Analysis (SNAP ID Technique): 1 µg/mL from a previous lot detected G9a (BAT8) on 10 µg of HeLa cell lysate.

Quality

Evaluated by Western Blot in HeLa cell lysate.
Western Blot Analysis: : 0.25-2 µg/mL of this antibody detected G9a (BAT8) in 10 µg of HeLa cell lysate.

Target description

~160 kDa

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Subbanna et al.
Neuroscience, 258, 422-432 (2013-12-05)
The transient exposure of immature rodents to ethanol during postnatal day 7 (P7), comparable to a time point within the third trimester of human pregnancy, induces neurodegeneration. However, the molecular mechanisms underlying the deleterious effects of ethanol on the developing
Shivakumar Subbanna et al.
Neurobiology of disease, 54, 475-485 (2013-02-12)
Rodent exposure to binge-like ethanol during postnatal day 7 (P7), which is comparable to the third trimester of human pregnancy, induces neuronal cell loss. However, the molecular mechanisms underlying these neuronal losses are still poorly understood. Here, we tested the
Sang Eun Park et al.
Oncotarget, 7(26), 39796-39808 (2016-10-26)
We previously reported that BIX-01294 (BIX), a small molecular inhibitor of euchromatic histone-lysine N-methyltransferase 2 (EHMT2/G9a), induces reactive oxygen species (ROS)-dependent autophagy in MCF-7 cells. Herein, we analyzed the epigenetic mechanism that regulates the transcription of Beclin-1, a tumor suppressor
Yongqing Liu et al.
Nature communications, 8(1), 355-355 (2017-08-27)
Impairment of intrinsic plasticity is involved in a range of neurological disorders such as epilepsy. However, how intrinsic excitability is regulated is still not fully understood. Here we report that the epigenetic factor Chromodomain Y-like (CDYL) protein is a critical
Yunha Kim et al.
Autophagy, 9(12), 2126-2139 (2013-12-11)
We screened a chemical library in MCF-7 cells stably expressing green fluorescent protein (GFP)-conjugated microtubule-associated protein 1 light chain 3 (LC3) (GFP-LC3-MCF-7) using cell-based assay, and identified BIX-01294 (BIX), a selective inhibitor of euchromatic histone-lysine N-methyltransferase 2 (EHMT2), as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service