Skip to Content
Merck
All Photos(4)

Key Documents

07-674

Sigma-Aldrich

Anti-Na+K+ ATPase α-2 Antibody

serum, Upstate®

Synonym(s):

Anti-Anti-DEE98, Anti-Anti-FARIMPD, Anti-Anti-FHM2, Anti-Anti-MHP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ATP1A2(477)

Specificity

Na+/K+ ATPase α-2
Predicted to crossreact with human, mouse, dog and monkey based on sequence homology.

Immunogen

KLH-conjugated, synthetic peptide (CKAGQENISVSKRDT) corresponding to amino acids 432-445 of human Na+K+ ATPase α-2, with an N-terminal cysteine added for conjugation purposes.

Application

Anti-Na+K+ ATPase α-2 Antibody is an antibody against Na+K+ ATPase α-2 for use in WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Quality

Routinely evaluated by immunoblot with rat brain microsomal preparation (catalog# 12-144)

Target description

Mr 105kDa

Physical form

Antiserum

Storage and Stability

Stable for 2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Taniguchi et al.
Journal of biochemistry, 129(3), 335-342 (2001-02-28)
Since the discovery of Na/K-ATPase, evidence has accumulated to suggest that 1 mol of ATP hydrolysis occurs via the Na(+)-occluded ADP-sensitive phosphoenzyme, the K(+)-sensitive phosphoenzyme and the K(+)-occluded enzyme accompanying active transport of 3Na(+) and 2K(+) according the Post-Albers scheme.
The molecular genetics of migraine.
Wessman, Maija, et al.
Annals of Medicine, 36, 462-473 (2004)
Tom Rouwette et al.
Molecular & cellular proteomics : MCP, 15(6), 2152-2168 (2016-04-23)
Chronic pain is a complex disease with limited treatment options. Several profiling efforts have been employed with the aim to dissect its molecular underpinnings. However, generated results are often inconsistent and nonoverlapping, which is largely because of inherent technical constraints.
The Na, K-ATPase in the failing human heart.
Schwinger, Robert H G, et al.
Cardiovascular Research, 57, 913-920 (2003)
Post-ischemic stimulation of 2-deoxyglucose uptake in rat myocardium: role of translocation of Glut-4
Montessuit, C, et al
Journal of Molecular and Cellular Cardiology, 30, 393-403 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service