Skip to Content
Merck
All Photos(2)

Key Documents

07-595

Sigma-Aldrich

Anti-acetyl-Histone H4 (Lys12) Antibody

serum, Upstate®

Synonym(s):

H4K12Ac, Histone H4 (acetyl K12)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

Saccharomyces cerevisiae, human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

acetylation (Lys12)

Gene Information

General description

Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.

Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).

Specificity

Broad species cross-reactivity expected
Recognizes Histone H4 acetylated on lysine 12.

Immunogen

peptide containing the sequence (LGAcKGG) corresponding to lysine 12 acetylation of yeast Histone H4.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-acetyl-Histone H4 (Lys12) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, DB, WB, ChIP-seq to detect acetyl-Histone H4 (Lys12) also known as H4K12Ac, Histone H4 (acetyl K12).

Quality

routinely evaluated by immunoblot on acid extracted proteins from HeLa cells

Target description

10 kDa

Physical form

Depleted rabbit antiserum in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.
Immunodepleted Serum

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Acid extracted proteins from HeLa cells treated with sodium butyrate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The molecular topography of silenced chromatin in Saccharomyces cerevisiae.
Thurtle, DM; Rine, J
Genes & Development null
Histone deacetylase inhibitors modify pancreatic cell fate determination and amplify endocrine progenitors.
Haumaitre, Cecile, et al.
Molecular and cellular biology, 28, 6373-6383 (2008)
The complex pattern of epigenomic variation between natural yeast strains at single-nucleosome resolution.
Filleton, F; Chuffart, F; Nagarajan, M; Bottin-Duplus, H; Yvert, G
Epigenetics & Chromatin null
Paola Y Bertucci et al.
Nucleic acids research, 41(12), 6072-6086 (2013-05-04)
Steroid receptors were classically described for regulating transcription by binding to target gene promoters. However, genome-wide studies reveal that steroid receptors-binding sites are mainly located at intragenic regions. To determine the role of these sites, we examined the effect of
Panagis Filippakopoulos et al.
Cell, 149(1), 214-231 (2012-04-03)
Bromodomains (BRDs) are protein interaction modules that specifically recognize ε-N-lysine acetylation motifs, a key event in the reading process of epigenetic marks. The 61 BRDs in the human genome cluster into eight families based on structure/sequence similarity. Here, we present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service