Skip to Content
Merck
All Photos(2)

Key Documents

07-488

Sigma-Aldrich

Anti-PKD2 Antibody

Upstate®, from rabbit

Synonym(s):

Serine/threonine-protein kinase D2, nPKC-D2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human
technique(s):
western blot: suitable
citations:
8

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PKD2(353503)

Specificity

Recognizes PKD2.

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 850-865 (GLPTDRDLGGACPPQD) of human protein kinase D2 (PKD2).

Application

Anti-PKD2 Antibody detects level of PKD2 & has been published & validated for use in WB.
Immunoblot Analysis: A 1:500-1:1,000 dilution of this antibody detected PKD2 in RIPA lysates from A431 cells that had been stimulated with PMA.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

Routinely evaluated by immunoblot in RIPA lysates from HEK293m cells that had been stimulated with 50 ng/mL of PDBu for 10 minutes or PMA stimulated A431 cells.

Target description

105 kDa

Physical form

ImmunoAffinity Purified
Immunoaffinity purified rabbit IgG in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl and 0.05% sodium azide as a preservative.

Storage and Stability

Maintain for 1 year at 2 to 8°C from date of shipment. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
PMA stimulated A431 cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein kinase d as a potential chemotherapeutic target for colorectal cancer.
Wei, N; Chu, E; Wipf, P; Schmitz, JC
Molecular Cancer Therapeutics null
Ying Wang et al.
The Journal of biological chemistry, 294(43), 15759-15767 (2019-09-08)
Vascular endothelial growth factor A (VEGF) signals primarily through its cognate receptor VEGF receptor-2 (VEGFR-2) to control vasculogenesis and angiogenesis, key physiological processes in cardiovascular disease and cancer. In human umbilical vein endothelial cells (HUVECs), knockdown of protein kinase D-1
Sabine Sturany et al.
The Journal of biological chemistry, 277(33), 29431-29436 (2002-06-12)
Recently, we cloned a novel serine/threonine kinase termed protein kinase D2 (PKD2). PKD2 can be activated by phorbol esters both in vivo and in vitro but also by gastrin via the cholecystokinin/CCK(B) receptor in human gastric cancer cells stably transfected
Nisha Durand et al.
Oncotarget, 9(91), 36358-36370 (2018-12-18)
Phosphatidylinositol-4-phosphate 5-kinase type-1C (PIP5K1C) is a lipid kinase that regulates focal adhesion dynamics and cell attachment through site-specific formation of phosphatidylinositol-4,5-bisphosphate (PI4,5P2). By comparing normal breast tissue to carcinoma in situ and invasive ductal carcinoma subtypes, we here show that
Ninel Azoitei et al.
Gut, 59(10), 1316-1330 (2010-08-25)
Tumour angiogenesis is crucially dependent on the communication between the tumour and the associated endothelium. Protein kinase D (PKD) isoenzymes mediate vascular endothelial growth factor-A (VEGF-A) induced endothelial cell proliferation and migration and are also highly expressed in various tumours.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service