07-1820
Anti-EEA1 Antibody
from rabbit, purified by affinity chromatography
Synonym(s):
early endosome antigen 1, early endosome-associated protein, early endosome antigen 1, 162kD, Endosome-associated protein p162, Zinc finger FYVE domain-containing protein 2
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
mouse, human
packaging
antibody small pack of 25 μL
technique(s)
immunocytochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... EEA1(8411)
General description
Specificity
Immunogen
Application
Signaling
General Post-translation Modification
Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected EEA1 in HeLa, A431, and C2C12 cells.
Quality
Western Blot Analysis: A 1:1000 dilution of this antibody detected EEA1 on 10 µg of BALB 3T3/A31 cell lysate.
Target description
Physical form
Storage and Stability
Analysis Note
BALB 3T3/A31 cell lysate
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service