Skip to Content
Merck
All Photos(2)

Key Documents

07-068-I

Sigma-Aldrich

Anti-Calcineurin A/PP2B catalytic unit Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Serine/threonine-protein phosphatase 2B catalytic subunit beta isoform, CAM-PRP catalytic subunit, Calmodulin-dependent calcineurin A subunit beta isoform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPP3CA(5530)
mouse ... Ppp3Ca(19055)
rat ... Ppp3Ca(24674)

General description

Calcineurin/PP2B Aβ, also known as Serine/threonine-protein phosphatase 2B catalytic subunit beta isoform, and encoded by the gene PPP3CB/CALNA2/CALNB/CNA2, is a calcium dependent, calmodulin stimulated catalytic subunit protein phosphatase critical in the function of Calcineurin an important phosphatase governing immune responses such as T cell activation and the T cell immune response. Calcineurin is a heterodimeric protein consisting of a catalytic subunit Calcineurin A, which contains an active site dinuclear metal center, and a tightly associated, myristoylated, Ca(2+)-binding subunit, Calcineurin B. Calcineurin/PP2B Aβ is the beta isoform of the catalytic active site of the subunit A of Calcineurin. Calcineurin also plays pivotal roles in signaling transduction cascades in a variety of other immune cells as well as non-immune cells like myocytes and neurons.

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to rat Calcineurin A/PP2B catalytic unit near the C-terminus.

Application

Immunoprecipitation Analysis: 5 µg from a representative lot immunoprecipitated Calcineurin A/PP2B catalytic unit in 500 µg of mouse brain tissue lysate.
Research Category
Signaling
Research Sub Category
Metabolic Hormones & Receptors
This Anti-Calcineurin A/PP2B catalytic unit antibody is validated for use in western blotting & IP for the detection of Calcineurin A/PP2B catalytic unit.

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Calcineurin A/PP2B catalytic unit in 10 µg of human brain tissue lysate.

Target description

~57 kDa observed

Linkage

Replaces: 07-068

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji-Eun Kim et al.
Frontiers in cellular neuroscience, 13, 179-179 (2019-05-24)
N-Methyl-D-aspartate receptor (NMDAR) and α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor (AMPAR) activations induce fast and transient mitochondrial fragmentation under pathophysiological conditions. However, it is still unknown whether NMDAR or AMPAR activity contributes to mitochondrial dynamics under physiological conditions. In the present study, MK801
Ji-Eun Kim et al.
Cells, 8(8) (2019-08-08)
2-Cyano-3,12-dioxo-oleana-1,9(11)-dien-28-oic acid methyl ester (CDDO-Me) is a triterpenoid analogue of oleanolic acid that exhibits promising anti-cancer, anti-inflammatory, antioxidant and neuroprotective activities. In addition, CDDO-Me affects cellular differentiation and cell cycle arrest, and irreversibly inhibits Lon protease-1 (LONP1). In the present
Ji-Eun Kim et al.
Biomedicines, 9(8) (2021-08-28)
The neural precursor cell expressed by developmentally downregulated gene 4-2 (NEDD4-2) is a ubiquitin E3 ligase that has a high affinity toward binding and ubiquitinating glutamate ionotropic receptor α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) type subunit 1 (GRIA1, also referred to GluR1 or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service