06-756
Anti-Frizzled 5 Antibody
Upstate®, from rabbit
Synonym(s):
Fz5, Fzd5
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
rat, human, mouse
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... FZD5(7855)
Related Categories
General description
The Wnt family consists of ligands involved in cell fate determination, organogenesis, and oncogenesis. The Wnt signal is thought to be transduced sequentially via a homolog of the frizzled family of receptors to GSK-3 (disheveled), β- and γ- catenin and finally to members of the Tcf/Lef-1 family of HMG box transcription factors. Frizzled 5 is believed to function as the receptor for the Wnt5a ligand.
Specificity
Frizzled 5 (aka Fz5 or Fzd5)
Immunogen
Epitope: C-Terminus
KLH-conjugated, synthetic peptide derived from amino acids 564-585 of human Frizzled 5 (CTGRTPPGPAATYHKQVSLSHV).
Application
Anti-Frizzled 5 Antibody is an antibody against Frizzled 5 for use in WB.
Research Category
Signaling
Signaling
Research Sub Category
Developmental Signaling
GPCR, cAMP/cGMP & Calcium Signaling
Developmental Signaling
GPCR, cAMP/cGMP & Calcium Signaling
Quality
Routinely evaluated by immunoblot on lysates from mouse liver, C2C12, Jurkat or HUVEC cells
Target description
Frizzled 5 (62kDa)
Physical form
Format: Purified
Protein A chromatography
Purified antibody in PBS with 0.05% sodium azide and 30% Glycerol
Storage and Stability
Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
Control
Lysates from mouse liver, C2C12, Jurkat, and HUVEC
Lysates from mouse liver, C2C12, Jurkat, and HUVEC
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Wnt/beta-catenin signaling pathway as a novel cancer drug target.
Current Cancer Drug Targets, 4, 653-671 (2004)
Norrin and frizzled; a new vein for the eye.
Developmental Cell, 6, 453-454 (2004)
Science (New York, N.Y.), 275(5306), 1652-1654 (1997-03-14)
In Xenopus laevis embryos, the Wingless/Wnt-1 subclass of Wnt molecules induces axis duplication, whereas the Wnt-5A subclass does not. This difference could be explained by distinct signal transduction pathways or by a lack of one or more Wnt-5A receptors during
The Journal of biological chemistry, 271(8), 4468-4476 (1996-02-23)
In Drosophila melanogaster, the frizzled gene plays an essential role in the development of tissue polarity as assessed by the orientation of cuticular structures. Through a combination of random cDNA sequencing, degenerate polymerase chain reaction amplification, and low stringency hybridization
The Journal of biological chemistry, 274(23), 16180-16187 (1999-05-29)
Frizzled related proteins (FRPs) comprise a family of secreted molecules that contain an N-terminal cysteine-rich domain (CRD) highly similar to the CRDs of the frizzled family of membrane-anchored Wnt receptors. FRPs have been shown to interact with Wnt proteins and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service