Skip to Content
Merck
All Photos(1)

Key Documents

06-747

Sigma-Aldrich

Anti-Neomycin Phosphotransferase II Antibody

Upstate®, from rabbit

Synonym(s):

Anti-Neomycin II, Neomycin II Antibody, Neomycin Phosphotransferase Ab

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, bacteria

species reactivity (predicted by homology)

mouse, rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Neomycin phosphotransferase II confers resistance to kanamycin and neomycin in bacteria and G418 (Geneticin®, G418 Sulfate) in mammalian cells. These antibiotics bind to ribosomal components and inhibit protein synthesis. NPT II inhibits these antibiotics through phosphorylation, which is thought to interfere with their active transport into the cell. Immunoblot analysis of transfected cell lysates with anti NPT II can be used to gauge relative transfection efficiencies in experiments where multiple transfections are performed in parallel with different plasmids.

Specificity

Recognizes NPT II, Mr ~30 kDa

Immunogen

Full length neomycin phosphotransferase (NPT) II

Application

Anti-Neomycin Phosphotransferase II Antibody is an antibody against Neomycin Phosphotransferase II for use in WB.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
Western Blot Analysis:
1-2 μg/mL dilutions of a previous lot detected NPT II in RIPA lysates from Cos-1 cells transfected with pUSE, harboring the neomycin resistance gene.

Quality

Routinely evaluated by Western Blot on transiently transfected Cos-1 cells.

Western Blot Analysis:
1-2 μg/mL of a previous lot detected NPT II in RIPA lysate from Cos-1 cells transfected with pUSE, harboring the neomycin resistance gene.

Target description

~30 kDa

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal IgG in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide before the addition of glycerol to 30%. Liquid at -20°C.

Storage and Stability

Stable for 1 year at 2 - 8°C from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at 2 - 8°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below 2 - 8°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
RIPA lysates from Cos-1 cells transfected with pUSE, harboring the neomycin resistance gene

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Geneticin is a registered trademark of Gibco BRL Life Technologies, Inc.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer E Dumaine et al.
eLife, 10 (2021-12-07)
The parasite Cryptosporidium is responsible for diarrheal disease in young children causing death, malnutrition, and growth delay. Cryptosporidium invades enterocytes where it develops in a unique intracellular niche. Infected cells exhibit profound changes in morphology, physiology, and transcriptional activity. How
An integrated strategy for efficient vector construction and multi-gene expression in Plasmodium falciparum.
Wagner, JC; Goldfless, SJ; Ganesan, SM; Lee, MC; Fidock, DA; Niles, JC
Malaria Journal null
Jan Kosla et al.
Cell communication and signaling : CCS, 11, 51-51 (2013-08-01)
Although there is extensive evidence for the amoeboid invasiveness of cancer cells in vitro, much less is known about the role of amoeboid invasiveness in metastasis and the importance of Rho/ROCK/MLC signaling in this process. We analyzed the dependence of
Efficient CRISPR-Cas9-mediated genome editing in Plasmodium falciparum.
Wagner, JC; Platt, RJ; Goldfless, SJ; Zhang, F; Niles, JC
Nature Methods null
Functional Characterization of ABCB4 Mutations Found in Low Phospholipid-Associated Cholelithiasis (LPAC).
Kim, TH; Park, HJ; Choi, JH
The Korean Journal of Physiology & Pharmacology : Official Journal of the Korean Physiological Society and the Korean Society of Pharmacology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service