Skip to Content
Merck
All Photos(4)

Key Documents

06-574

Sigma-Aldrich

Anti-TrkA Antibody

Upstate®, from rabbit

Synonym(s):

Neurotrophic tyrosine kinase receptor type, Oncogene TRK, TRK1-transforming tyrosine kinase protein, high affinity nerve growth factor receptor, neurotrophic tyrosine kinase, receptor, type 1, tyrosine kinase receptor A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

TrkA, apparent molecular weight 140 kDa, is a high affinity nerve growth factor (NGF). The Trk proto-oncogene family contains four members, TrkA, TrkB, TrkC, and TrkE, which are variably expressed throughout the central and peripheral nervous systems. TrkA binds to nerve growth factor (NGF) and autophosphorylates on tyrosine residues (Tyr490, Tyr674, Tyr675, Tyr751 and Tyr785) to activate multiple downstream effector proteins. Phosphorylation at Tyr490 is required for Shc association and subsequent activation of the Ras-MAP kinase-signaling cascade, which leads to activation of Elk-1-dependent gene transcription and neurite growth. Phosphorylations at Tyr674 and Tyr675 lie within the catalytic domain of TrkA tyrosine kinase and reflect Trk kinase activity. Additionally, phosphorylation at Tyr751 is required for PI3-kinase association and activation of the Akt signaling cascade.

Specificity

Reactivity with other species has not been confirmed.
Recognizes TrkA, Mr 140 kDa. Does not cross-react with TrkB or TrkC.

Immunogen

Rabbits immunized with bacterially expressed purified protein corresponding to the entire extracellular domain of rat TrkA receptor.

Application

Anti-TrkA Antibody detects level of TrkA & has been published & validated for use in IC, IP & WB.
Immunoprecipitation:
4 μg of a previous lot immunoprecipitated TrkA from 500 μg of PC-12 RIPA lysate.

Immunocytochemistry:
10 μg/mL of a previous lot of this antibody showed positive immunostaining for TrkA in PC-12 cells fixed with 4% paraformaldehyde.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neurodegenerative Diseases

Quality

Routinely evaluated by Western Blot on RIPA lysates from human A431 cells, mouse 3T3 or rat PC-12 cell lysates.

Western Blot Analysis:
0.5-2 μg/mL of this lot detected TrkA in RIPA lysates from human A431 cells; a previous lot detected TrkA in mouse 3T3 and rat PC-12 cell lysates.

Target description

140 kDa

Linkage

Replaces: CBL770

Physical form

Format: Purified
Protein A purified
Purified rabbit IgG in buffer containing 0.02 M phosphate buffer, pH 7.6, 0.25 M NaCl, 0.1% sodium azide. Frozen at -20°C.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerolcontaining solutions to become frozen during storage.

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An alternatively spliced form of the nerve growth factor receptor TrkA confers an enhanced response to neurotrophin 3
Clary, D. O. and Reichardt, L. F.
Proceedings of the National Academy of Sciences of the USA, 91, 11133-11137 (1994)
The Gata3 transcription factor is required for the survival of embryonic and adult sympathetic neurons.
Tsarovina, K; Reiff, T; Stubbusch, J; Kurek, D; Grosveld, FG; Parlato, R; Schutz, G; Rohrer, H
The Journal of Neuroscience null
p21-activated protein kinase 4 (PAK4) interacts with the keratinocyte growth factor receptor and participates in keratinocyte growth factor-mediated inhibition of oxidant-induced cell death.
Lu, Y; Pan, ZZ; Devaux, Y; Ray, P
The Journal of Biological Chemistry null
Establishment of neurovascular congruency in the mouse whisker system by an independent patterning mechanism.
Oh, WJ; Gu, C
Neuron null
Rui Ji et al.
PloS one, 9(12), e115140-e115140 (2014-12-17)
Tyro3, Axl and Mertk (TAM) receptor tyrosine kinases play multiple functional roles by either providing intrinsic trophic support for cell growth or regulating the expression of target genes that are important in the homeostatic regulation of immune responses. TAM receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service