Skip to Content
Merck
All Photos(7)

Key Documents

05-368

Sigma-Aldrich

Anti-phospho-Ser/Thr-Pro MPM-2 Antibody

clone MPM-2, Upstate®, from mouse

Synonym(s):

Phospho-Ser/Thr Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

MPM-2, monoclonal

species reactivity

vertebrates

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

phosphorylation (pSer/pThr)

General description

This antibody recognizes a variety of proteins that are phosphorylated during mitosis.
Upon entry into M (Mitosis) phase, many proteins are phosphorylated either directly or indirectly by M-phase-promoting factor (MPF). The MPM2 monoclonal antibody binds to a phospho amino acid-containing epitope (peptides containing LTPLK and FTPLQ domains) present on more than 50 proteins of M-phase eukaryotic cells.

Specificity

Recognizes phosphorylated serines or threonines when followed by a proline.

Immunogen

Mitotic human HeLa cell cytosolic lysate

Application

Anti-phospho-Ser/Thr-Pro MPM-2 Antibody is a Mouse Monoclonal Antibody for detection of phospho-Ser/Thr-Pro also known as Mitotic protein #2 & has been tested in ELISA, IHC, IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

routinely evaluated in immunoblot on colcemid treated human HeLa carcinoma cells

Target description

varies depending on the protein you are detecting

Physical form

Format: Purified
Protein G Purified
Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Colcemid treated HeLa cell lysate or A431 mitotic cells fixed with 3.7% paraformaldehyde

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibitor-2 induced M-phase arrest in Xenopus cycling egg extracts is dependent on MAPK activation.
Arian Khandani,Mahmood Mohtashami,Anne Camirand
Cellular & Molecular Biology Letters null
J Cobb et al.
Developmental biology, 205(1), 49-64 (1999-01-12)
Little is known about the timing of meiotic prophase events during spermatogenesis in the mouse or how these events are related to cell-cycle progression. This work was designed to test hypotheses about the timing and biochemical correlates of developmental acquisition
S A Innocente et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(5), 2147-2152 (1999-03-03)
The p53 tumor suppressor controls multiple cell cycle checkpoints regulating the mammalian response to DNA damage. To identify the mechanism by which p53 regulates G2, we have derived a human ovarian cell that undergoes p53-dependent G2 arrest at 32 degrees
Human papillomavirus type 16 E7 oncoprotein engages but does not abrogate the mitotic spindle assembly checkpoint.
Yu, Y; Munger, K
Virology null
Cyclin E controls Drosophila female germline stem cell maintenance independently of its role in proliferation by modulating responsiveness to niche signals.
Ables, ET; Drummond-Barbosa, D
Development null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service