Skip to Content
Merck
All Photos(1)

Documents

05-321X

Sigma-Aldrich

Anti-Phosphotyrosine Antibody, clone 4G10®

clone 4G10®, Upstate®, from mouse

Synonym(s):

Clone 4G10 Anti-Phosphotyrosine, Phosphotyrosine Detection Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G10®, monoclonal

species reactivity

vertebrates

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr)

Related Categories

General description

Some of the tyrosine residues can be tagged with a phosphate group (phosphorylated) by protein kinases. (In its phosphorylated state, it is referred to as phosphotyrosine.). Tyrosine phosphorylation is considered as one of the key steps in signal transduction and regulation of enzymatic activity.
The advent of anti-phosphotyrosine antibodies is one of significant events in signal transduction research. Before the availability of anti-phosphotyrosine antibodies, tyrosyl phospyhorylation of proteins and enzymes was investigated through hazardous and time-consuming radioactive experiments. Anti-phosphotyrosine antibodies are commonly used in western blots after the targeted proteins have been immunoprecipitated to measure the tyrosyl phosphorylation of the proteins. Anti-phosphotyrosine antibodies are also directly used on cell lysate to examine the overall change of tyrosine phosphorylation level in reponse to various treatments.

Specificity

This antibody recognizes tyrosine-phosphorylated proteins from all species.

Immunogen

Phosphotyramine-KLH

Application

Anti-Phosphotyrosine Antibody, clone 4G10 detects tyrosine phosphorylated proteins in all species. This unique monoclonal antibody is validated for use in IC, IH, IP, WB and is backed by hundreds of publications.
Immunoprecipitation:
2-4 μg of a previous lot immunoprecipitated quantitatively the phosphotyrosine containing proteins in the lysate of a confluent culture (10 cm dish) of cells expressing an activated tyrosine kinase. To preserve phosphotyrosine, add: 0.2 mM sodium orthovanadate to the lysis buffer.
Research Category
Signaling
Research Sub Category
General Post-translation Modification

Quality

Routinely evaluated on EGF-treated human A431 carcinoma cells.

Western Blot Analysis:
0.5-2 μg/mL of this lot detected tyrosine-phosphorylated proteins in a modified RIPA lysate from EGF-treated human A431 carcinoma cells (Cohen, B., 1990; Druker, B. J.,1989; Kanakura, Y., 1991).

Target description

Dependent upon the molecular weight of the tyrosine phosphorylated protein being detected.

Physical form

Format: Purified
Protein G-Sepharose Chromatography
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide. Liquid at 2-8ºC.
IgG2bκ mouse monoclonal antibody produced in vitro by mouse-mouse hybridoma 4G10 (FOX-NY [NS-1 derivative] myeloma x spleen cells).

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

NOTE: DO NOT FREEZE.
For maximum recovery of the product, centrifuge the original vial prior to removing the cap. If the product has accidentally been frozen and thawed, spin it at 13,000 x g for 10 minutes at 2-8ºC. Save the supernatant for application.

Analysis Note

Control
Positive Antigen Control: Catalog #12-302, EGF-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ALK-activating homologous mutations in LTK induce cellular transformation.
Roll, JD; Reuther, GW
Testing null
Chi Yan et al.
Frontiers in molecular biosciences, 9, 730213-730213 (2022-06-21)
Background: Target therapies play more and more important roles in gastrointestinal stromal tumors (GISTs) and melanoma with the advancement of clinical drugs that overcome the resistance caused by gene mutations. c-KIT gene mutations account for a large portion of GIST
Polo kinase links the stress pathway to cell cycle control and tip growth in fission yeast.
Janni Petersen, Iain M Hagan
Nature null
E Josué Ruiz et al.
Molecular cell, 32(2), 210-220 (2008-10-28)
Cell-cycle progression is regulated by cyclin-dependent kinases (CDKs). CDK1 and CDK2 can be also activated by noncyclin proteins named RINGO/Speedy, which were identified as inducers of the G2/M transition in Xenopus oocytes. However, it is unclear how XRINGO triggers M
Qin Xia et al.
iScience, 24(12), 103528-103528 (2021-12-18)
Amplification of ubiquitin E3 ligase Smurf1 promotes degradation of PTEN leading to hyperactivation of the Akt/mTORC1 pathway. However, inhibitors of this pathway have not hitherto yielded promising results in clinical studies because of strong drug resistance. Here, we investigated Smurf1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service