Skip to Content
Merck
All Photos(1)

Key Documents

05-172

Sigma-Aldrich

Anti-IGF-I Antibody, clone Sm1.2

clone Sm1.2, Upstate®, from mouse

Synonym(s):

Mechano growth factor, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), somatomedin C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

Sm1.2, monoclonal

species reactivity

avian, chicken, mouse, rat, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
neutralization: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... IGF1(3479)

General description

Insulin-like growth factor 1 (IGF-1) is a polypeptide protein hormone similar in molecular structure to insulin. It plays an important role in childhood growth and continues to have anabolic effects in adults. IGF-1 consists of 70 amino acids in a single chain with three intramolecular disulfide bridges.
IGF-1 is produced primarily by the liver as an endocrine hormone and targets tissues in a paracrine/autocrine fashion. Production is stimulated by growth hormone and can be retarded by undernutrition, growth hormone insensitivity, lack of growth hormone receptors, or failures of the downstream signaling pathway post GH receptor including SHP2 and STAT5b. Approximately 98% of IGF-1 is always bound to one of 6 binding proteins (IGF-BP). IGFBP-3, the most abundant protein, accounts for 80% of all IGF binding. IGF-1 binds to IGFBP-3 in a 1:1 molar ratio. Its primary action is mediated by binding to specific IGF receptors present on many cell types in many tissues. The signal is transduced by intracellular events. IGF-1 is one of the most potent natural activators of the AKT signaling pathway, a stimulator of cell growth and multiplication and a potent inhibitor of programmed cell death.

Specificity

IGF-I. (Russel, W.E., 1984). 40% cross reactivity to IGF-II. (van Wyk, J.J., 1997).
Possibly chicken. Occasional difficulty neutralizing rat IGF-I due to binding protein interference.

Immunogen

Human insulin-like growth factor I (IGF-I) purified from human plasma. (van Wyk, J.J., 1986).

Application

Anti-IGF-I Antibody, clone Sm1.2 is an antibody against IGF-I for use in IH, IP, NEUT & WB.
Immunoprecipitation:
5 μg of a previous lot immunoprecipitated 100 ng of IGF-I.

Immunohistochemistry:
10 μg/mL of a previous lot was reported by an independent laboratory to detect IFG-I in formalin-fixed, paraffin-embedded skin sections.
(Lui, 1994).

Neutralization:
10-20 μg/mL of a previous lot inhibited the activity of 10 ng/mL of IGF-I as determined by testing with chicken embryo fibroblasts (CEF) using an ATP endpoint assay (ATPLiteTM-M, Packard Instruments).
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Quality

Evaluated on IGF-I (Catalog # 01-208) under non-reducing conditions.

Western Blot Analysis:
0.5-2 μg/mL of this antibody detected 100 ng of IGF-I (Catalog # 01-208) under non-reducing conditions.

Target description

17 kDa, migrates to ~7 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse IgG1κ lyophilized powder, lyophilized from 105 μL of 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.1 mM EDTA.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
MCF-7 cell extracts

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PTH ameliorates acidosis-induced adverse effects in skeletal growth centers: the PTH-IGF-I axis.
Green, J; Goldberg, R; Maor, G
Kidney International null
Inhibition of the mitogenic effects of plasma by a monoclonal antibody to somatomedin C.
Russell, W E, et al.
Proceedings of the National Academy of Sciences of the USA, 81, 2389-2392 (1984)
G Hudelist et al.
Endocrine-related cancer, 14(4), 1053-1062 (2007-11-30)
BRCA1/2 mutations predispose to early onset breast and ovarian cancers. The phenotypic expression of mutant alleles, however, is thought to be modified by factors that are also involved in the pathogenesis of sporadic breast cancer. One such protein is IGF-I
Sex hormone-induced prostatic carcinogenesis in the noble rat: the role of insulin-like growth factor-I (IGF-I) and vascular endothelial growth factor (VEGF) in the development of prostate cancer.
Y Z Wang, Y C Wong
Prostate null
Expression of insulin-like growth factor I (IGF-I) in nevi and melanomas.
M G Fleming, S F Howe, L H Graf
The American Journal of Dermatopathology null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service