Skip to Content
Merck
All Photos(1)

Key Documents

04-902

Sigma-Aldrich

Anti-HA Tag Antibody, clone DW2, rabbit monoclonal

culture supernatant, clone DW2, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

DW2, monoclonal

species reactivity

vertebrates (common species)

technique(s)

western blot: suitable

isotype

IgG

shipped in

dry ice

target post-translational modification

unmodified

General description

Human influenza hemagglutinin (HA) is a surface glycoprotein required for the infectivity of the human virus. The HA tag is derived from the HA molecule that corresponds to amino acids 98-106. It has been extensively used as a general epitope tag in expression vectors. Many recombinant proteins have been engineered to express the HA tag, which does not appear to interfere with the bioactivity or the biodistribution of the recombinant protein. This tag facilitates the detection, isolation, and purification of the proteins.

Specificity

Reactivity with other species has not been confirmed.
Recognizes recombinant proteins containing the HA epitope tag. The apparent molecular weight is related to the tagged protein.

Immunogen

KLH-conjugated, synthetic peptide containing the sequence …CYPYDVPDY… derived from the influenza virus hemaglutinin (HA) protein.

Application

Anti-HA Tag Antibody, clone DW2 is an antibody against HA Tag for use in WB.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
Western Blot Analysis: A 1:1,000-1:2,000 dilution of this lot detected HA-tagged WNT-1 protein expressed in HEK293 cells.

Quality

Western Blot Analysis
Lysates from non-transfected (lane 1) and HAtagged WNT-1 transfected (lane 2) HEK293 cells were resolved by electrophoresis, transferred to nitrocellulose and probed with anti-HA-Tag, clone DW2 (1:1,000 dilution).
Proteins were visualized using a goat anti-rabbit secondary antibody conjugated to HRP and a chemiluminescence detection system.

Arrow indicates the HA-tagged protein (~49 kDa). The apparent molecular weight is related to the tagged protein.

Target description

Various

Linkage

Replaces: 05-902

Physical form

Cultured supernatant in 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of shipment. Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HA-tagged WNT-1 protein.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivonne Melano et al.
International journal of molecular sciences, 25(1) (2024-01-11)
How ACE2 functions as the major host receptor of SARS-CoV-2 despite having low expression in the lungs is still unknown. To facilitate the development of therapeutic strategies against coronaviruses, gaining a deeper comprehension of the molecular mechanism of SARS-CoV-2 infection
Elinn Johansson et al.
Cell reports, 20(7), 1641-1653 (2017-08-17)
Hypoxia-inducible factors enhance glioma stemness, and glioma stem cells have an amplified hypoxic response despite residing within a perivascular niche. Still, little is known about differential HIF regulation in stem versus bulk glioma cells. We show that the intracellular domain
Yuag-Meng Liu et al.
Journal of biomedical science, 26(1), 58-58 (2019-08-17)
Multiple interplays between viral and host factors are involved in influenza virus replication and pathogenesis. Several small RNAs have recently emerged as important regulators of host response to viral infections. The aim of this study was to characterize the functional
Saikat Bhattacharya et al.
Nature communications, 12(1), 6452-6452 (2021-11-10)
The RNA recognition motif (RRM) binds to nucleic acids as well as proteins. More than one such domain is found in the pre-mRNA processing hnRNP proteins. While the mode of RNA recognition by RRMs is known, the molecular basis of
Wen-Chi Su et al.
Journal of virology, 92(4) (2017-11-24)
Virus replication is mediated by interactions between the virus and host. Here, we demonstrate that influenza A virus membrane protein 2 (M2) can be ubiquitinated. The lysine residue at position 78, which is located in the cytoplasmic domain of M2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service