Skip to Content
Merck
All Photos(1)

Key Documents

Z-902

Supelco

Zopiclone-D4 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13ClN6O3D4
CAS Number:
Molecular Weight:
392.83
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C1C2=NC=CN=C2C(OC(N3C([2H])([2H])CN(C)CC3([2H])[2H])=O)N1C4=NC=C(Cl)C=C4

InChI

1S/C17H17ClN6O3/c1-22-6-8-23(9-7-22)17(26)27-16-14-13(19-4-5-20-14)15(25)24(16)12-3-2-11(18)10-21-12/h2-5,10,16H,6-9H2,1H3/i8D2,9D2

InChI key

GBBSUAFBMRNDJC-LZMSFWOYSA-N

General description

This internal standard is suitable for many LC/MS and GC/MS applications including forensic analysis, clinical toxicology, urine drug testing, or isotope dilution methods. Zopiclone is a non-benzodiazepine hypnotic sold under the trade names Imovane® and Zimovane and is prescribed for the treatment of insomnia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Imovane is a registered trademark of Sanofi-Aventis
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth Beer
Journal of drugs in dermatology : JDD, 11(12), 1494-1495 (2013-02-05)
Eruptions from drugs are among the most frequently reported adverse events. The most frequent etiologic agents include antibiotics and nonsteroidal anti-inflammatory agents. These eruptions are also a frequent source of visits to dermatology of!ces. Many of the inciting agents are
[New Levitra in the treatment of erectile dysfunction].
A L Vertkin et al.
Urologiia (Moscow, Russia : 1999), (6)(6), 100-102 (2013-02-06)
Christoph Zenzmaier et al.
Endocrinology, 153(11), 5546-5555 (2012-09-06)
Phosphodiesterase type 5 (PDE5) inhibitors have been demonstrated to improve lower urinary tract symptoms secondary to benign prostatic hyperplasia (BPH). Because BPH is primarily driven by fibroblast-to-myofibroblast trans-differentiation, this study aimed to evaluate the potential of the PDE5 inhibitor vardenafil
Daniel J Mans et al.
Journal of pharmaceutical and biomedical analysis, 75, 153-157 (2012-12-25)
Ion mobility spectrometry was used as a rapid screening tool for the detection of acetildenafils, sildenafils and avanafil within adulterated herbal supplement matrices. Acetildenafils show a tendency for partial fragmentation during the desorption/ionization process affording two peaks in the ion
S I Gamidov et al.
Urologiia (Moscow, Russia : 1999), (3)(3), 102-104 (2013-08-31)
Due to high efficiency, prompt action and low incidence of side effects, phosphodiesterase type 5 inhibitors are the drugs of first choice in the treatment of erectile dysfunction (ED). One of the most widely used drugs in this group currently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service