Skip to Content
Merck
All Photos(1)

Key Documents

T-080

Supelco

(±)-cis-11-Nor-9-carboxy-Δ9-THC-D3 glucuronide solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)-cis-11-Nor-9-carboxyΔ9-THC-d3 glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H33O10D3
Molecular Weight:
523.59
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])CCCCc1cc(O)c2C3C=C(CCC3C(C)(C)Oc2c1)C(=O)O[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(O)=O

InChI

1S/C27H36O10/c1-4-5-6-7-13-10-17(28)19-15-12-14(8-9-16(15)27(2,3)37-18(19)11-13)25(34)36-26-22(31)20(29)21(30)23(35-26)24(32)33/h10-12,15-16,20-23,26,28-31H,4-9H2,1-3H3,(H,32,33)/t15?,16?,20-,21-,22+,23-,26-/m0/s1/i1D3

InChI key

UWXZAZRSQQLWKH-IOHBEHRKSA-N

General description

A stable-labeled internal standard for 11-Nor-9-carboxy-Δ9-THC glucuronide, a major urinary metabolite of Δ9-THC. This Certified Spiking Solution® is suitable for LC/MS or GC/MS applications such as forensic testing, toxicology analysis, urine drug testing, or isotope dilution methods. This internal standard provides added value in allowing for quantitation of glucuronide levels by LC-MS/MS without need of an added hydrolysis step.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacqueline A Hubbard et al.
Clinical chemistry and laboratory medicine, 58(5), 673-681 (2019-09-19)
Background The widespread availability of cannabis raises concerns regarding its effect on driving performance and operation of complex equipment. Currently, there are no established safe driving limits regarding ∆9-tetrahydrocannabinol (THC) concentrations in blood or breath. Daily cannabis users build up

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service