Skip to Content
Merck
All Photos(1)

Key Documents

T-002

Supelco

Trinitroglycerin solution

1000 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5N3O9
CAS Number:
Molecular Weight:
227.09
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1000 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

O=[N+]([O-])OCC(CO[N+]([O-])=O)O[N+]([O-])=O

InChI

1S/C3H5N3O9/c7-4(8)13-1-3(15-6(11)12)2-14-5(9)10/h3H,1-2H2

InChI key

SNIOPGDIGTZGOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trinitroglycerin, also commonly known as nitroglycerin, is a pharmaceutical vasodilator used to treat heart conditions such as angina and chronic heart failure. This explosive compound is also an ingredient in dynamite and gunpowder. This Certified Spiking Solution® is suitable for use in chromatography or mass spectrometry-based applications in pharmaceutical research or clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hosein Kimiaei Asadi et al.
Acta cirurgica brasileira, 28(1), 19-25 (2013-01-23)
To evaluate the effect of nitroglycerine (NTG) on sensory and motor block onset and recovery time as well as the quality of tourniquet pain relief, when added to lidocaine (LID) for intravenous regional anesthesia in elective forearm and hand surgery.
Huarong Xu et al.
Journal of mass spectrometry : JMS, 48(4), 519-532 (2013-04-16)
A novel, sensitive and reliable ultra fast liquid chromatography-tandem mass spectrometry (UFLC-MS/MS) method has been developed and validated for simultaneous quantitation of eight main active ingredients (evodiamine, rutaecarpine, dehydroevodiamine, limonin, ginsenoside Rb1, Rd, Re and Rg1) in rat plasma after
Kate Kleydman et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 38(12), 1889-1897 (2012-12-05)
Skin necrosis after soft tissue augmentation with dermal fillers is a rare but potentially severe complication. Nitroglycerin paste may be an important treatment option for dermal and epidermal ischemia in cosmetic surgery. To summarize the knowledge about nitroglycerin paste in
Mikhail Kosiborod et al.
Journal of the American College of Cardiology, 61(20), 2038-2045 (2013-03-19)
This study sought to examine the efficacy of ranolazine versus placebo on weekly angina frequency and sublingual nitroglycerin use in subjects with type 2 diabetes mellitus, coronary artery disease (CAD), and chronic stable angina who remain symptomatic despite treatment with
Guillaume Ruel et al.
Nutrition research (New York, N.Y.), 33(1), 41-49 (2013-01-29)
The stiffening of arteries is a key step in atherogenesis leading to cardiovascular disease. It has been suggested that dietary polyphenols may be cardioprotective through possible favorable effects on oxidative stress and vascular function. The present study was undertaken in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service