Skip to Content
Merck
All Photos(1)

Key Documents

S-008

Supelco

Sufentanil Citrate solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N2O2S · C6H8O7
CAS Number:
Molecular Weight:
578.67
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

-10 to -25°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCC(=O)N(c1ccccc1)C2(CCN(CCc3cccs3)CC2)COC

InChI

1S/C22H30N2O2S.C6H8O7/c1-3-21(25)24(19-8-5-4-6-9-19)22(18-26-2)12-15-23(16-13-22)14-11-20-10-7-17-27-20;7-3(8)1-6(13,5(11)12)2-4(9)10/h4-10,17H,3,11-16,18H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

OJCZPLDERGDQRJ-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

General description

Sufentanil is a short acting synthetic opioid analgesic most commonly used for pain relief during medical operations. This certified solution standard is applicable to use in clinical toxicology, pain prescription monitoring, or forensic analysis by LC-MS/MS or GC/MS. Sufentanil is sold under the trade name Sufenta.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Xin Wang et al.
Knee surgery, sports traumatology, arthroscopy : official journal of the ESSKA, 21(5), 1140-1145 (2012-12-12)
To evaluate the effect of combined local anaesthetic and opioid in post-operative analgesia, the effect of intra-articular injection of local anaesthetic (ropivacaine), opioid (sufentanil) and combination of these two (ropivacaine combined with sufentanil) after the single-bundle anterior cruciate ligament reconstruction
J Fechner et al.
European journal of pain (London, England), 17(4), 562-570 (2012-08-29)
There is an ongoing debate whether opioids when used for intra-operative analgesia may enhance post-operative pain. We studied the effect of two different intra-operative dosings of sufentanil on post-operative morphine consumption, pain and hyperalgesia after cardiac anaesthesia. Forty-two male patients
Robert Stephen et al.
Journal of opioid management, 8(4), 237-241 (2012-09-04)
In emergency medicine, the ability to provide rapid, adequate pain control without high resource utilization is ideal. In this study, the efficacy of intranasal sufentanil in emergency department (ED) patients with acute distal extremity injury was evaluated. A nonrandomized, open-label
S Karason et al.
Acta anaesthesiologica Scandinavica, 57(5), 639-645 (2013-02-26)
Bladder catheterisation may be inconvenient for patients, delay mobilisation and risk complications. We hypothesised that by excluding pre-operatively patients at high risk of post-operative urinary retention, the majority of patients could avoid perioperative catheterisation during low-dose spinal anaesthesia. Patients undergoing
Marc Lilot et al.
Anesthesia and analgesia, 117(1), 259-264 (2013-04-18)
In this randomized, prospective trial, we sought to determine the effective dose of hypobaric ropivacaine with sufentanil providing 95% success (ED95) in spinal anesthesia for traumatic femoral neck surgery in the elderly. Sixty-eight elderly patients with unilateral hip fracture randomly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service