Skip to Content
Merck
All Photos(1)

Key Documents

P-063

Supelco

Phenytoin solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C1NC(=O)C(N1)(c2ccccc2)c3ccccc3

InChI

1S/C15H12N2O2/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,(H2,16,17,18,19)

InChI key

CXOFVDLJLONNDW-UHFFFAOYSA-N

General description

Phenytoin is an antiepileptic drug used in the prevention and treatment of seizures. This analytical reference standard is suitable as starting material for the preparation of calibrators and controls in phenytoin testing by GC/MS or LC-MS/MS. Phenytoin is sold under the trade name Dilantin®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Dilantin is a registered trademark of Warner-Lambert Co.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bradley R Keel et al.
Tennessee medicine : journal of the Tennessee Medical Association, 106(4), 35-36 (2013-04-27)
Anticonvulsant Hypersensitivity Syndrome (AHS) is a rare complication of common drugs used today. It is unusual in that it occurs later than most other drug reactions, about two to six weeks after initiation of the offending agent. It also has
Elisa Colombo et al.
PloS one, 8(1), e55329-e55329 (2013-02-06)
The persistent Na⁺ current (I(NaP)) is important for neuronal functions and can play a role in several pathologies, although it is small compared to the transient Na⁺ current (I(NaT)). Notably, I(NaP) is not a real persistent current because it undergoes
B Medhi et al.
Indian journal of physiology and pharmacology, 56(4), 382-387 (2013-06-21)
Esomeprazole is commonly prescribed proton pump inhibitor for gastritis and peptic ulcer disease. Most of the time in clinical practice, phenytoin and esomeprazole are prescribed for patients of generalized seizures with concomitant peptic ulcer. Hence there are chances of drug-drug
Kimford J Meador et al.
The Lancet. Neurology, 12(3), 244-252 (2013-01-29)
Many women of childbearing potential take antiepileptic drugs, but the cognitive effects of fetal exposure are uncertain. We aimed to assess effects of commonly used antiepileptic drugs on cognitive outcomes in children up to 6 years of age. In this
Amitava Dasgupta et al.
Journal of clinical laboratory analysis, 27(1), 1-4 (2013-01-18)
For many years, fluorescence polarization immunoassay (FPIA) on the TDx analyzer has been used for determination of free phenytoin concentration. Recently Abbott Laboratories decided to discontinue the TDx analyzer and related assays on this analyzer. Free phenytoin assay is also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service