Skip to Content
Merck
All Photos(1)

Documents

N-904

Supelco

(±)-Norpropoxyphene-d5 maleate solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D5H22NO2 · C4H4O4
CAS Number:
Molecular Weight:
446.55
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(Cc2ccccc2)(OC(=O)CC)C(C)CNC

InChI

1S/C21H27NO2.C4H4O4/c1-4-20(23)24-21(17(2)16-22-3,19-13-9-6-10-14-19)15-18-11-7-5-8-12-18;5-3(6)1-2-4(7)8/h5-14,17,22H,4,15-16H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/i6D,9D,10D,13D,14D;

InChI key

HCQPFYNZJNOOKN-ZXYHGPIJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Norpropoxyphene is a major urinary metabolite of propoxyphene, an opioid analgesic drug used for the treatment of pain. Norpropoxyphene′s toxicity makes the parent drug more likely to cause death from overdose compared to similar mild opioid analgesics. Suitable use of this stable-labeled internal standard includes quantitation of norpropoxyphene levels by GC/MS or LC/MS in testing applications such as clinical toxicology, urine drug testing, isotope dilution methods or pain prescription monitoring.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service