Skip to Content
Merck
All Photos(1)

Key Documents

M-912

Supelco

Methandienone solution

1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in 1,2-dimethoxyethane

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C20H28O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h6,9,12,15-17,22H,4-5,7-8,10-11H2,1-3H3/t15-,16+,17+,18+,19+,20+/m1/s1

InChI key

XWALNWXLMVGSFR-HLXURNFRSA-N

Gene Information

human ... AR(367)

General description

A Certified Spiking Solution® applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as sports testing, urine drug testing, clinical toxicology, and forensic analysis. Methandrostenolone, also called methandienone, is an anabolic steroid sold under the trade name Dianabol®. This performance enhancing drug has an extensive history of abuse by bodybuilders.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Dianabol is a registered trademark of Jared R. Wheat
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cholestasis secondary to anabolic steroid use in young men.
Ahmed M Elsharkawy et al.
BMJ (Clinical research ed.), 344, e468-e468 (2012-02-04)
Leen Lootens et al.
Drug testing and analysis, 1(11-12), 531-537 (2010-04-01)
Anabolic androgenic steroids are considered to be doping agents and are prohibited in sports. Their metabolism needs to be elucidated to allow for urinary detection by gas chromatography-mass spectrometry (GC-MS) or liquid chromatography-tandem mass spectrometry (LC-MS/MS). Steroid metabolism was assessed
Yun Wang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(22), 2149-2154 (2011-06-28)
Methandrostenolone (MA) is a steroid used as veterinary medicine on stockbreeding to promote animal growth. The use of MA has been strictly regulated because of its harmful effect on consumers. This paper describes the production of polyclonal antibody (pAb) against
Beth A Costine et al.
Psychoneuroendocrinology, 35(10), 1473-1485 (2010-06-12)
In the past several decades, the therapeutic use of anabolic androgenic steroids (AAS) has been overshadowed by illicit use of these drugs by elite athletes and a growing number of adolescents to enhance performance and body image. As with adults
Stephanie Mosler et al.
Archives of toxicology, 86(1), 109-119 (2011-08-06)
There is increasing evidence that the biological activity of myostatin (MSTN), a negative regulator of muscle growth, is affected by training but also anabolic steroids. In this study, we analyzed the effects of the frequently abused anabolic steroid methandienone (Md)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service