Skip to Content
Merck
All Photos(1)

Key Documents

F-003

Supelco

Flurazepam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23ClFN3O
CAS Number:
Molecular Weight:
387.88
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

O=C1CN=C(C2=CC=CC=C2F)C3=CC(Cl)=CC=C3N1CCN(CC)CC

InChI

1S/C21H23ClFN3O.2ClH/c1-3-25(4-2)11-12-26-19-10-9-15(22)13-17(19)21(24-14-20(26)27)16-7-5-6-8-18(16)23;;/h5-10,13H,3-4,11-12,14H2,1-2H3;2*1H

InChI key

ZIIJJOPLRSCQNX-UHFFFAOYSA-N

General description

Flurazepam is a benzodiazepine, sold under the trade names Dalmane and Salmadorm for treament of mild to moderate insomnia. This analytical reference standard is suitable for many LC/MS or GC/MS applications from urine drug testing and clinical toxicology to forensic toxicology analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michal Abrahamowicz et al.
Statistics in medicine, 31(11-12), 1014-1030 (2011-11-19)
Pharmacoepidemiology investigates associations between time-varying medication use/dose and risk of adverse events. Applied research typically relies on a priori chosen simple conventional models, such as current dose or any use in the past 3 months. However, different models imply different risk
Cristian Palmiere et al.
Forensic science international, 188(1-3), e17-e19 (2009-05-05)
A case of sustained combustion of a human body that occurred in 2006 in Geneva, Switzerland, is presented. The body of a man was discovered at home and found to have been almost completely incinerated between the knees and the
Jessica A Burket et al.
Epilepsy & behavior : E&B, 16(3), 415-417 (2009-09-19)
The inbred Balb/c mouse strain was more sensitive than the outbred NIH Swiss mouse to flurazepam's ability to antagonize electrically precipitated seizures. In prior work, a reduction in flurazepam's antiseizure efficacy was not observed 24h after forcing Balb/c mice to
Damien E Earl et al.
Neural plasticity, 2012, 405926-405926 (2012-07-26)
Cessation of one-week oral administration of the benzodiazepine flurazepam (FZP) to rats results in withdrawal anxiety after 1 day of withdrawal. FZP withdrawal is correlated with synaptic incorporation of homomeric GluA1-containing α-amino-3-hydroxy-5-methylisoxazole-4-propionic acid receptors (AMPARs) in the proximal stratum radiatum
Susan M Hanson et al.
British journal of pharmacology, 162(3), 673-687 (2010-10-15)
Although the functional effects of benzodiazepines (BZDs) on GABA(A) receptors have been well characterized, the structural mechanism by which these modulators alter activation of the receptor by GABA is still undefined. We used disulphide trapping between engineered cysteines to probe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service