Skip to Content
Merck
All Photos(1)

Key Documents

ERR-001S

Supelco

RDX solution

1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®

Synonym(s):

Hexahydro-1,3,5-trinitro-1,3,5-triazine, Hexahydro-1,3,5-trinitro-1,3,5-triazine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N6O6
CAS Number:
Molecular Weight:
222.12
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

1000 μg/mL in acetonitrile

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

O=[N+]([O-])N1CN([N+]([O-])=O)CN([N+]([O-])=O)C1

InChI

1S/C3H6N6O6/c10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15/h1-3H2

InChI key

XTFIVUDBNACUBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J G Giarrizzo et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 925, 70-75 (2013-03-26)
A simple, fast liquid-liquid extraction method was developed for studying hexahydro-1,3,5-trinitro-1,3,5 triazine (RDX) biodegradation using small sample volumes. The method was tested in vitro with anaerobic incubations of RDX with whole rumen fluid (WRF) and a commercial Sporanaerobacter acetigenes strain
Karl J Indest et al.
Applied and environmental microbiology, 79(5), 1746-1750 (2013-01-01)
The transcriptome of RDX (hexahydro-1,3,5-trinitro-1,3,5-triazine)-degrading strain Gordonia sp. strain KTR9 and its glnR mutant were studied as a function of nitrogen availability to further investigate the observed ammonium-mediated inhibition of RDX degradation. The results indicate that nitrogen availability is a
Anat Bernstein et al.
Environmental science & technology, 47(1), 479-484 (2012-12-12)
The explosive Hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX) is known to be degraded aerobically by various isolates of the Rhodococcus species, with denitration being the key step, mediated by Cytochrome P450. Our study aimed at gaining insight into the RDX degradation mechanism by Rhodococcus
Karl K Irikura
The journal of physical chemistry. A, 117(10), 2233-2241 (2013-02-05)
The explosive nitramine RDX (1,3,5-trinitrohexahydro-s-triazine) is thought to decompose largely by homolytic N-N bond cleavage, among other possible initiation reactions. Density-functional theory (DFT) calculations indicate that the resulting secondary aminyl (R2N·) radical can abstract an oxygen atom from NO2 or
Hao-Ping Chen et al.
Applied and environmental microbiology, 78(21), 7798-7800 (2012-08-28)
Whole-genome sequencing, transcriptomic analyses, and metabolic reconstruction were used to investigate Gordonia sp. strain KTR9's ability to catabolize a range of compounds, including explosives and steroids. Aspects of this mycolic acid-containing actinobacterium's catabolic potential were experimentally verified and compared with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service