Skip to Content
Merck
All Photos(1)

Key Documents

E-068

Supelco

Everolimus solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H83NO14
CAS Number:
Molecular Weight:
958.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

shipped in

dry ice

storage temp.

−70°C

SMILES string

N21[C@@H](CCCC2)C(=O)O[C@@H](CC(=O)[C@@H](\C=C(\[C@H]([C@H](C(=O)[C@@H](C[C@@H](\C=C\C=C\C=C(\[C@H](C[C@H]4O[C@]([C@@H](CC4)C)(C(=O)C1=O)O)OC)/C)C)C)OC)O)/C)C)[C@@H](C[C@H]3C[C@H]([C@@H](CC3)OCCO)OC)C

InChI

1S/C53H83NO14/c1-32-16-12-11-13-17-33(2)44(63-8)30-40-21-19-38(7)53(62,68-40)50(59)51(60)54-23-15-14-18-41(54)52(61)67-45(35(4)28-39-20-22-43(66-25-24-55)46(29-39)64-9)31-42(56)34(3)27-37(6)48(58)49(65-10)47(57)36(5)26-32/h11-13,16-17,27,32,34-36,38-41,43-46,48-49,55,58,62H,14-15,18-26,28-31H2,1-10H3/b13-11+,16-12+,33-17+,37-27+/t32-,34-,35-,36-,38-,39+,40+,41+,43-,44+,45+,46-,48-,49+,53-/m1/s1

InChI key

HKVAMNSJSFKALM-GKUWKFKPSA-N

Gene Information

human ... FKBP1A(2280)

General description

Everolimus, sold under trade names including Zortress®, Certican, and Afinitor®, is an immunosuppressant drug used to prevent rejection of organ transplants and to treat renal cell cancer and other tumors. This Certified Spiking Solution® is suitable as starting material for calibrators, controls, or linearity standards for therapeutic drug monitoring or clinical and diagnostic testing of everolimus in patient whole blood samples by LC-MS/MS.

Legal Information

Afinitor is a registered trademark of Novartis AG
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Zortress is a registered trademark of Novartis AG

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matteo Santoni et al.
Biochimica et biophysica acta, 1845(2), 221-231 (2014-02-01)
The mammalian target of rapamycin (mTOR) has emerged as an attractive cancer therapeutic target. Treatment of metastatic renal cell carcinoma (mRCC) has improved significantly with the advent of agents targeting the mTOR pathway, such as temsirolimus and everolimus. Unfortunately, a
Eleonora Teplinsky et al.
Breast cancer research and treatment, 141(2), 167-172 (2013-09-05)
Hepatitis B reactivation can occur with cytotoxic chemotherapy in patients with hepatitis B and cancer. Reactivation can occur in a patient with chronic hepatitis, an inactive carrier, or one with resolved hepatitis. Clinical presentation may range from subclinical elevation of
Michael Cardamone et al.
The Journal of pediatrics, 164(5), 1195-1200 (2014-02-13)
To evaluate the efficacy and side effects of oral mammalian target of rapamycin (mTOR) inhibitors in children and adolescents with tuberous sclerosis complex (TSC) and intractable epilepsy or subependymal giant cell astrocytoma (SEGA). Single-center series of 13 children and adolescents
Elisavet Paplomata et al.
Breast cancer research and treatment, 140(3), 453-462 (2013-08-03)
Everolimus is an orally available inhibitor of the mammalian target of rapamycin (mTOR), which has been approved in combination with exemestane for hormone receptor-positive (HR) breast cancer after failure of treatment with non-steroidal aromatase inhibitors. Everolimus is generally very well
Everolimus is a potent inhibitor of activated hepatic stellate cell functions in vitro and in vivo, while demonstrating anti-angiogenic activities.
Piguet AC, Majumder S, Maheshwari U, et al.
Clinical Science, 126(11), 775-784 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service