Skip to Content
Merck
All Photos(1)

Key Documents

D-916

Supelco

N-Desmethylclomipramine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21ClN2 · HCl
CAS Number:
Molecular Weight:
337.29
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC2=C(C=C1)CCC3=C(C=CC=C3)N2CCCNC.[H]Cl

InChI

1S/C18H21ClN2.ClH/c1-20-11-4-12-21-17-6-3-2-5-14(17)7-8-15-9-10-16(19)13-18(15)21;/h2-3,5-6,9-10,13,20H,4,7-8,11-12H2,1H3;1H

InChI key

KMDDAZOLOSKTKZ-UHFFFAOYSA-N

General description

N-Desmethylclomipramine is a primary plasma metabolite of the tricyclic antidepressant clomipramine. Clomipramine is a tricyclic antidepressant used to treat many conditions from major depression and panic disorder to narcolepsy and obsessive compulsion disorder (OCD). This Certified Snap-N-Spike® Solution is applicable for use in urine drug testing, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Dale et al.
Veterinary and human toxicology, 36(4), 309-310 (1994-08-01)
A 27-y-old male was admitted deeply comatosed 5-6 h after taking approximately 15 g clomipramine. The prominent feature of the case was a biphasic course of clomipramine and desmethylclomipramine serum concentrations, possibly caused by delayed drug absorption. Clinically, 2 serious
Clinical pharmacology and therapeutics, 66(2), 152-165 (1999-08-25)
To examine the problems of establishing dose-effect and concentration-effect relationships of antidepressant therapy with clomipramine. This randomized double-blind study compared five fixed doses of clomipramine hydrochloride: 25, 50, 75, 125, and 200 mg/day in hospitalized or day patients at nine
F Coudoré et al.
Journal of analytical toxicology, 20(2), 101-105 (1996-03-01)
A rapid and efficient high-performance liquid chromatographic method using a reversed-phase eluent of methanol-water with butylamine on a silica column was developed for the separation and quantitation of clomipramine (CMI), its demethylated metabolites (desmethyl-clomipramine and didesmethyl-clomipramine [DDCMI]), and its hydroxylated
Plasma concentrations after a clomipramine intoxication.
L M Stolk et al.
Journal of analytical toxicology, 22(7), 612-613 (1998-12-10)
K Shimoda et al.
Journal of clinical psychopharmacology, 19(5), 393-400 (1999-10-03)
The aim of this study was to compare the disposition of the tricyclic antidepressant clomipramine (C) in Japanese and Swedish patients receiving continuous treatment. Therapeutic drug monitoring data for C and the active metabolite N-desmethylclomipramine (DC) in Japanese patients receiving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service