Skip to Content
Merck
All Photos(1)

Key Documents

D-062

Supelco

21-Deoxycortisol solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O4
CAS Number:
Molecular Weight:
346.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CC(=O)C1(O)CCC2C3CCC4=CC(=O)CCC4(C)C3C(O)CC12C

InChI

1S/C21H30O4/c1-12(22)21(25)9-7-16-15-5-4-13-10-14(23)6-8-19(13,2)18(15)17(24)11-20(16,21)3/h10,15-18,24-25H,4-9,11H2,1-3H3/t15-,16-,17-,18+,19-,20-,21-/m0/s1

InChI key

LCZBQMKVFQNSJR-UJPCIWJBSA-N

General description

A new calibration material suitable for LC/MS applications in clinical and diagnostic testing and endocrinology. 21-deoxycortisol, a steroid intermediate in the formation of cortisol, is measured in whole blood or serum by LC/MS for newborn screening of congenital adrenal hyperplasia (CAH) and other adrenal diseases.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vânia Tonetto-Fernandes et al.
Journal of pediatric endocrinology & metabolism : JPEM, 21(5), 487-494 (2008-07-29)
21-Hydroxylase deficiency (21OHD) is the commonest form of congenital adrenal hyperplasia, while 11betaOHD represents 5% of cases. Although both result from mutations in distinct genes, cases of 'apparent' combined 21OHD and 11betaOHD (AC21,11OHD) have been occasionally reported. A 6 year-old
M Hill et al.
Steroids, 60(9), 615-620 (1995-09-01)
A radioimmunoassay of three deoxycorticoids, namely 11 beta,17 alpha-dihydroxy-4-pregnene-3,20-dione (21-deoxycortisol), 17 alpha,21-dihydroxy-4-pregnene-3,20-dione (11-deoxycortisol), and 21-hydroxy-4-pregnene-3,20-dione (11-deoxycorticosterone) which are important for differential diagnosis of congenital adrenal disorders, is described and evaluated. Antisera against 3-(O-carboxymethyl)oximes conjugated to bovine serum albumin were raised
Surface-activated chemical ionization ion trap mass spectrometry in the analysis of 21-deoxycortisol in blood.
Simone Cristoni et al.
Rapid communications in mass spectrometry : RCM, 18(12), 1392-1396 (2004-06-03)
J Fiet et al.
Annals of clinical biochemistry, 31 ( Pt 1), 56-64 (1994-01-01)
21-deoxycortisol (21-DF) is a steroid of strictly adrenal origin formed by the 11-hydroxylation of 17-hydroxyprogesterone. This metabolic pathway is minor in normal subjects, in whom basal plasma concentrations range from 0.03 to 0.63 nmol/L and from 0.865 to 1.50 nmol/L
N S Sieber-Ruckstuhl et al.
Domestic animal endocrinology, 31(1), 63-75 (2005-10-20)
Trilostane is thought to be a competitive inhibitor of the 3beta-hydroxysteroid dehydrogenase (3beta-HSD), an essential enzyme system for the synthesis of cortisol, aldosterone and androstenedione. Due to its reliable clinical efficacy, trilostane is increasingly used to treat dogs with pituitary-dependant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service