Skip to Content
Merck
All Photos(1)

Key Documents

D-049

Supelco

N-Desmethylclobazam solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClN2O2
CAS Number:
Molecular Weight:
286.71
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1)N(C2=CC=CC=C2)C3=C(C=CC(Cl)=C3)NC1=O

InChI

1S/C15H11ClN2O2/c16-10-6-7-12-13(8-10)18(11-4-2-1-3-5-11)15(20)9-14(19)17-12/h1-8H,9H2,(H,17,19)

InChI key

RRTVVRIFVKKTJK-UHFFFAOYSA-N

General description

A Certified Solution Standard for use in LC or GC applications from forensic and forensic toxicology analysis to urine drug testing. N-Desmethylclobazam, found in urine, serum, and plasma, is the pharmacologically active, major metabolite of clobazam.

Application



  • Impact on bioavailability in epilepsy treatment: A study discussed the implications of N-Desmethylclobazam on drug to drug interactions and neuropsychological impact when used as add-on therapy in drug-resistant epilepsies, highlighting its role in enhancing therapeutic bioavailability (Pietrafusa et al., 2023).


  • Research on pharmacokinetic variability: Research explored the pharmacokinetic variability of N-Desmethylclobazam, emphasizing its influence by age, drug interactions, and biochemical markers of toxicity in patients with epilepsy, contributing to a deeper understanding of its pharmacological properties (Heger et al., 2024).


  • Evaluation of drug-drug interactions: A comprehensive analysis investigated potential drug-drug interactions between N-Desmethylclobazam and anti-seizure medications, providing insights into its safe and effective use in clinical settings (Gaston et al., 2023).


  • Synergistic effects in epilepsy management: A multicenter study on the cenobamate-clobazam interaction revealed both pharmacokinetic interactions and evidence of synergy, underscoring N-Desmethylclobazam′s potential to enhance treatment outcomes in epilepsy (Osborn and Abou-Khalil, 2023).


  • Analysis of metabolites in forensic science: Research on the measurement of benzodiazepine receptor agonists′ metabolites, including N-Desmethylclobazam, outlined its detection and concentration levels in urine, offering applications in forensic toxicology (Volkova et al., 2023).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Herráez-Hernández et al.
Journal of pharmaceutical and biomedical analysis, 14(8-10), 1077-1087 (1996-06-01)
An on-line dialysis-solid-phase extraction-gas chromatographic (GC) approach has been developed for the determination of drugs in plasma, using some benzodiazepines as model compounds. Clean-up is based on performing the dialysis of 100 microliters samples for 7 min using water as
F Nakamura et al.
Epilepsia, 37(8), 728-735 (1996-08-01)
The antiepileptic effects of clobazam, a 1,5-benzodiazepine, have been well documented in animal experiments and clinical trials. However, the drug's mechanisms of antiepileptic actions are still undetermined. The purpose of this study was to learn how clobazam and its active
K K Akerman
Scandinavian journal of clinical and laboratory investigation, 56(7), 609-614 (1996-11-01)
This report describes a simple reversed-phase high-performance liquid chromatographic (HPLC) method with automated solid-phase extraction (SPE) for analysing clobazam and norclobazam concentrations in human serum or plasma. For the HPLC analysis the samples and standards are prepared with an ASPEC
T Pullar et al.
British journal of clinical pharmacology, 23(3), 317-321 (1987-03-01)
The effect of cimetidine on the single dose pharmacokinetics of orally administered clobazam and N-desmethylclobazam (NDMC) was studied in volunteers. Cimetidine inhibited the elimination of both clobazam and NDMC and inhibited the rate of formation of NDMC from clobazam. The
D F LeGatt et al.
Clinical biochemistry, 26(3), 159-163 (1993-06-01)
We have developed a simple, rapid method for determination of the antiepileptic drug clobazam and its major active metabolite, norclobazam in serum. Serum (200 microL) made alkaline with sodium borate buffer is extracted with toluene. After evaporation of the organic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service