Skip to Content
Merck
All Photos(1)

Documents

B-001

Supelco

Benzoylecgonine-d3 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Benzoylecgonine-N-methyl-d3 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D3H16NO4
CAS Number:
Molecular Weight:
292.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

mass shift

M+3

shipped in

wet ice

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])N1[C@H]2CC[C@@H]1[C@H]([C@H](C2)OC(=O)c3ccccc3)C(O)=O

InChI

1S/C16H19NO4/c1-17-11-7-8-12(17)14(15(18)19)13(9-11)21-16(20)10-5-3-2-4-6-10/h2-6,11-14H,7-9H2,1H3,(H,18,19)/t11-,12+,13-,14+/m0/s1/i1D3

InChI key

GVGYEFKIHJTNQZ-GZBWLTEMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard suitable for isotope dilution methods or benzoylecgonine testing in urine, saliva, blood, or hair by GC/MS or LC/MS for applications in clinical toxicology, urine drug testing, pain prescription monitoring or forensic analysis. Benzoylecgonine is the primary metabolite of cocaine and a major biomarker for determination of cocaine use.

Application


  • Mass Spectrometry Internal Standard: Benzoylecgonine-d₃ is utilized as a robust internal standard in mass spectrometry to enhance the accuracy and precision of quantitative analysis of cocaine metabolites in clinical toxicology studies, providing crucial data for forensic and clinical applications (Handley et al., 2019).

  • Forensic Toxicology Research: The application of Benzoylecgonine-d₃ in gas chromatography-mass spectrometry (GC-MS) underscores its value in the forensic analysis of cocaine and its metabolites in biological samples, offering reliable quantification essential for legal and medical investigations (Fleming et al., 2010).

  • LC-MS/MS Quantitative Analysis: Benzoylecgonine-d₃ serves as a critical component in developing validated LC-MS/MS methodologies for drug testing, ensuring the traceability and reproducibility required in doping control and therapeutic drug monitoring (Lin et al., 2001).

  • Pharmacokinetic Studies: The use of stable isotope-labeled Benzoylecgonine-d₃ enables detailed pharmacokinetic studies, helping to understand the metabolism of cocaine and assess the impact of various physiological and pathological conditions on drug metabolism (Lin et al., 2001).

  • Drug Metabolism Analysis: In forensic science, Benzoylecgonine-d₃ is crucial for developing sensitive and selective analytical methods for detecting and quantifying cocaine use, supporting both clinical diagnostics and legal proceedings (Hall et al., 1999).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James Brian McAlvin et al.
Investigative ophthalmology & visual science, 56(6), 3820-3826 (2015-06-13)
Amino-amide or amino-ester local anesthetics, which are currently used for topical ocular anesthesia, are short acting and may delay corneal healing with long-term use. In contrast, site 1 sodium channel blockers (S1SCBs) are potent local anesthetics with minimal adverse tissue
Paula E Park et al.
Addiction biology, 20(2), 275-284 (2013-12-18)
Opioids represent effective drugs for the relief of pain, yet chronic opioid use often leads to a state of increased sensitivity to pain that is exacerbated during withdrawal. A sensitization of pain-related negative affect has been hypothesized to closely interact
Kalpashri Kesavan et al.
Respiratory physiology & neurobiology, 201, 93-100 (2014-07-11)
Sedative-analgesics are often given to newborn infants and are known to affect many components of the autonomic nervous system. While morphine is most frequently used, α-2 adrenergic receptor agonists are being increasingly used in this population. Alpha-2 adrenergic receptors agonists
Craige C Wrenn et al.
Neuroscience letters, 585, 109-113 (2014-11-30)
Fragile X syndrome (FXS), a disorder caused by a mutation in the FMR1 gene, is often associated with Attention Deficit Hyperactivity Disorder (ADHD). Common treatments for the hyperactivity often seen in ADHD involve the use of stimulants and α2-adrenergic agonists.
Oruc Allahverdiyev et al.
Turkish journal of medical sciences, 45(6), 1380-1389 (2016-01-19)
Treating animals repeatedly with intermittent and increasing morphine doses has been suggested to allow some withdrawal during each dosing interval, which causes repeated stress. The present study aimed to test this hypothesis and assess sex-related differences in withdrawal signs and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service